Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578401_m1
          See other NRP2 GE Assays ›
          Gene Symbol:
          Nrp2
          Gene Name:
          neuropilin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 69496878 - 69609802 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578401_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030869.3 NP_110496.1 5 - 6 1100 86
          XM_006245039.2 XP_006245101.1 5 - 6 1658 86
          XM_006245040.2 XP_006245102.1 5 - 6 1657 86
          XM_006245041.2 XP_006245103.1 5 - 6 1659 86
          XM_006245042.2 XP_006245104.1 5 - 6 1658 86
          XM_008767137.1 XP_008765359.1 5 - 6 1657 86
          GenBank mRNA AF016297.1 - 5 - 6 1133 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000782.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nrp2

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          neural crest cell migration
          neuron migration
          axon guidance
          heart development
          facial nerve structural organization
          vestibulocochlear nerve structural organization
          nerve development
          gonadotrophin-releasing hormone neuronal migration to the hypothalamus
          ventral trunk neural crest cell migration
          vascular endothelial growth factor signaling pathway
          axon extension involved in axon guidance
          negative chemotaxis
          sympathetic ganglion development
          trigeminal ganglion development
          ganglion morphogenesis
          semaphorin-plexin signaling pathway
          sensory neuron axon guidance
          sympathetic neuron projection extension
          sympathetic neuron projection guidance
          neural crest cell migration involved in autonomic nervous system development
          semaphorin-plexin signaling pathway involved in neuron projection guidance
          facioacoustic ganglion development
          vascular endothelial growth factor-activated receptor activity
          heparin binding
          semaphorin receptor activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129840 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60251 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178362 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42843 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12465 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12465 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12465 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15555 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1972 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76841 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58755 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35728 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17372 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923842 - -
          Affymetrix Rat Gene 1.0 ST Array 10923843 - -
          Affymetrix Rat Gene 1.0 ST Array 10923840 - -
          Affymetrix Rat Gene 1.0 ST Array 10923841 - -
          Affymetrix Rat Gene 1.0 ST Array 10923853 - -
          Affymetrix Rat Gene 1.0 ST Array 10923850 - -
          Affymetrix Rat Gene 1.0 ST Array 10923845 - -
          Affymetrix Rat Gene 1.0 ST Array 10923848 - -
          Affymetrix Rat Gene 1.0 ST Array 10923837 - -
          Affymetrix Rat Gene 1.0 ST Array 10923847 - -
          Affymetrix Rat Gene 1.0 ST Array 10923849 - -
          Affymetrix Rat Gene 1.0 ST Array 10923852 - -
          Affymetrix Rat Gene 1.0 ST Array 10923854 - -
          Affymetrix Rat Gene 1.0 ST Array 10923844 - -
          Affymetrix Rat Gene 1.0 ST Array 10923846 - -
          Affymetrix Rat Gene 1.0 ST Array 10923838 - -
          Affymetrix Rat Gene 1.0 ST Array 10923836 - -
          Affymetrix Rat Gene 1.0 ST Array 10923839 - -
          Affymetrix Rat Gene 1.1 ST Array 10923835 - -
          Rat Genome 230 2.0 Array 1369824_at - -
          Rat Genome 230 2.0 Array 1369824_PM_at - -
          Affymetrix HT Rat Focus Array 1369824_at - -
          Affymetrix HT Rat Focus Array 1369824_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900006323.rn.1 TC0900000782.rn.1 RTA TC
          Rat Expression Set 230 1369824_PM_at - -
          Rat Expression Set 230 1369824_at - -
          Affymetrix Rat Gene 1.0 ST Array 10923835 - -
          Rat Genome U34 Set AF016297_at - -
          Rat Genome U34 Set AF016297_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22369114 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040866 - -
          RatRef-12 Expression BeadChip ILMN_62549 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline