Search
Search
| Gene Symbol: Akr1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031000.3 | NP_112262.1 | 2 - 3 | 136 | 136 | |
| GenBank mRNA | BC059133.1 | - | 2 - 3 | 415 | 6918695 | 136 |
| D10854.1 | - | 2 - 3 | 136 | 136 | ||
| FQ212779.1 | - | 2 - 3 | 411 | 136 | ||
| FQ212919.1 | - | 1 - 2 | 86 | 136 | ||
| FQ213220.1 | - | 1 - 2 | 83 | 136 | ||
| FQ213467.1 | - | 2 - 3 | 411 | 136 | ||
| FQ213483.1 | - | 2 - 3 | 407 | 136 | ||
| FQ213579.1 | - | 2 - 3 | 378 | 136 | ||
| FQ213776.1 | - | 2 - 3 | 378 | 136 | ||
| FQ214173.1 | - | 2 - 3 | 378 | 136 | ||
| FQ214700.1 | - | 2 - 3 | 351 | 136 | ||
| FQ215620.1 | - | 2 - 3 | 203 | 136 | ||
| FQ215772.1 | - | 2 - 3 | 378 | 136 | ||
| FQ218383.1 | - | 2 - 3 | 378 | 136 | ||
| FQ218721.1 | - | 2 - 3 | 378 | 136 | ||
| FQ226221.1 | - | 2 - 3 | 411 | 136 | ||
| FQ228131.1 | - | 2 - 3 | 411 | 136 | ||
| FQ228132.1 | - | 2 - 3 | 411 | 136 | ||
| FQ228842.1 | - | 2 - 3 | 407 | 136 | ||
| FQ233156.1 | - | 2 - 3 | 411 | 136 | ||
Set Membership: |
|