Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578778_m1
          See other ABCC6 GE Assays ›
          Gene Symbol:
          Abcc6
          Gene Name:
          ATP-binding cassette, subfamily C (CFTR/MRP), member 6
          Gene Aliases:
          Mrp6
          Chromosome Location:
          Chr.1: 101959513 - 102013243 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578778_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcc6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031013.1 NP_112275.1 10 - 11 1402 83
          XM_006229159.2 XP_006229221.1 10 - 11 1319 83
          XM_008759419.1 XP_008757641.1 9 - 10 1648 83
          GenBank mRNA AB010466.1 - 10 - 11 1355 83
          U73038.1 - 10 - 11 1402 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005415.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abcc6

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          transmembrane transport
          ATP binding
          ATPase activity
          ATPase activity, coupled to transmembrane movement of substances

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P463998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163037 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59417 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13451 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P463998 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P463998 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P463998 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P463998 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700936086 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60853 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93794 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10368 - -
          Agilent Rat cDNA Microarray (G4105A) 700936086 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10721982 - -
          Affymetrix Rat Gene 1.0 ST Array 10721993 - -
          Affymetrix Rat Gene 1.0 ST Array 10721973 - -
          Affymetrix Rat Gene 1.0 ST Array 10721980 - -
          Affymetrix Rat Gene 1.0 ST Array 10721974 - -
          Affymetrix Rat Gene 1.0 ST Array 10721986 - -
          Affymetrix Rat Gene 1.0 ST Array 10721989 - -
          Affymetrix Rat Gene 1.0 ST Array 10721977 - -
          Affymetrix Rat Gene 1.0 ST Array 10721981 - -
          Affymetrix Rat Gene 1.0 ST Array 10721994 - -
          Affymetrix Rat Gene 1.0 ST Array 10721979 - -
          Affymetrix Rat Gene 1.0 ST Array 10721999 - -
          Affymetrix Rat Gene 1.0 ST Array 10721975 - -
          Affymetrix Rat Gene 1.0 ST Array 10721984 - -
          Affymetrix Rat Gene 1.0 ST Array 10721978 - -
          Affymetrix Rat Gene 1.0 ST Array 10721998 - -
          Affymetrix Rat Gene 1.0 ST Array 10721988 - -
          Affymetrix Rat Gene 1.0 ST Array 10721996 - -
          Affymetrix Rat Gene 1.0 ST Array 10721991 - -
          Affymetrix Rat Gene 1.0 ST Array 10721976 - -
          Affymetrix Rat Gene 1.0 ST Array 10721985 - -
          Affymetrix Rat Gene 1.0 ST Array 10721983 - -
          Affymetrix Rat Gene 1.0 ST Array 10721987 - -
          Affymetrix Rat Gene 1.0 ST Array 10721995 - -
          Affymetrix Rat Gene 1.0 ST Array 10721970 - -
          Affymetrix Rat Gene 1.0 ST Array 10721990 - -
          Affymetrix Rat Gene 1.0 ST Array 10721969 - -
          Affymetrix Rat Gene 1.0 ST Array 10721992 - -
          Affymetrix Rat Gene 1.0 ST Array 10721971 - -
          Affymetrix Rat Gene 1.0 ST Array 10721972 - -
          Affymetrix Rat Gene 1.1 ST Array 10721968 - -
          Rat Genome 230 2.0 Array 1368452_at - -
          Rat Genome 230 2.0 Array 1368452_PM_at - -
          Affymetrix HT Rat Focus Array 1368452_at - -
          Affymetrix HT Rat Focus Array 1368452_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100044344.rn.1 TC0100005415.rn.1 RTA TC
          Rat Expression Set 230 1368452_PM_at - -
          Rat Expression Set 230 1368452_at - -
          Affymetrix Rat Gene 1.0 ST Array 10721968 - -
          Rat Genome U34 Set AB010466_s_at - -
          Rat Genome U34 Set AB010466_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22125919 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361474 - -
          RatRef-12 Expression BeadChip ILMN_53785 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline