Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578842_m1
          See other MAPK14 GE Assays ›
          Gene Symbol:
          Mapk14
          Gene Name:
          mitogen activated protein kinase 14
          Gene Aliases:
          CRK1, CSBP, CSPB1, Csbp1, Csbp2, Exip, Hog, Mxi2, Prkm14, Prkm15, RK, Sapk2A, p38, p38Hog, p38alpha
          Chromosome Location:
          Chr.20: 5933303 - 5995137 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578842_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mapk14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031020.2 NP_112282.2 1 - 2 419 87
          XM_006256142.2 XP_006256204.1 1 - 2 131 87
          GenBank mRNA AB000783.1 - 1 - 2 418 87
          AF346293.1 - 1 - 2 116 87
          BC092193.1 - 1 - 2 419 7313217 87
          U73142.1 - 1 - 2 127 87
          U91847.1 - 1 - 2 116 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000234.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mapk14

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          MAPK cascade
          cell morphogenesis
          cartilage condensation
          angiogenesis
          placenta development
          chondrocyte differentiation
          glucose metabolic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          protein phosphorylation
          apoptotic process
          cellular response to DNA damage stimulus
          transmembrane receptor protein serine/threonine kinase signaling pathway
          skeletal muscle tissue development
          response to glucose
          positive regulation of gene expression
          positive regulation of myotube differentiation
          myoblast differentiation involved in skeletal muscle regeneration
          peptidyl-serine phosphorylation
          fatty acid oxidation
          osteoclast differentiation
          lipopolysaccharide-mediated signaling pathway
          response to muramyl dipeptide
          response to lipopolysaccharide
          intracellular signal transduction
          cellular response to vascular endothelial growth factor stimulus
          response to muscle stretch
          p38MAPK cascade
          positive regulation of protein import into nucleus
          signal transduction in response to DNA damage
          positive regulation of blood vessel endothelial cell migration
          positive regulation of erythrocyte differentiation
          positive regulation of myoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of glucose import
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          striated muscle cell differentiation
          stress-activated MAPK cascade
          positive regulation of cardiac muscle cell proliferation
          cellular response to lipopolysaccharide
          cellular response to ionizing radiation
          negative regulation of canonical Wnt signaling pathway
          positive regulation of brown fat cell differentiation
          stress-induced premature senescence
          positive regulation of myoblast fusion
          positive regulation of reactive oxygen species metabolic process
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase activity
          protein binding
          ATP binding
          protein C-terminus binding
          kinase activity
          NFAT protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P258485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20515 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P258485 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P258485 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P258485 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P286911 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12479 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P360409 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701911766 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63319 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53448 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85522 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46513 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91696 - -
          Agilent Rat cDNA Microarray (G4105A) 701911766 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828765 - -
          Affymetrix Rat Gene 1.0 ST Array 10828774 - -
          Affymetrix Rat Gene 1.0 ST Array 10828768 - -
          Affymetrix Rat Gene 1.0 ST Array 10828771 - -
          Affymetrix Rat Gene 1.0 ST Array 10828769 - -
          Affymetrix Rat Gene 1.0 ST Array 10828770 - -
          Affymetrix Rat Gene 1.0 ST Array 10828764 - -
          Affymetrix Rat Gene 1.0 ST Array 10828767 - -
          Affymetrix Rat Gene 1.0 ST Array 10828763 - -
          Affymetrix Rat Gene 1.0 ST Array 10828777 - -
          Affymetrix Rat Gene 1.0 ST Array 10828776 - -
          Affymetrix Rat Gene 1.0 ST Array 10828773 - -
          Affymetrix Rat Gene 1.0 ST Array 10828766 - -
          Affymetrix Rat Gene 1.0 ST Array 10828775 - -
          Affymetrix Rat Gene 1.1 ST Array 10828762 - -
          Rat Genome 230 2.0 Array 1367697_PM_at - -
          Rat Genome 230 2.0 Array 1367697_at - -
          Affymetrix HT Rat Focus Array 1367697_PM_at - -
          Affymetrix HT Rat Focus Array 1367697_at - -
          Affymetrix Rat Clariom S Assay PSR2000002302.rn.1 TC2000000234.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000002301.rn.1 TC2000000234.rn.1 RTA TC
          Rat Expression Set 230 1367697_at - -
          Rat Expression Set 230 1367697_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828762 - -
          Rat Genome U34 Set U73142_at - -
          Rat Genome U34 Set U91847_s_at - -
          Rat Genome U34 Set rc_AI171630_s_at - -
          Rat Genome U34 Set rc_AI137862_s_at - -
          Rat Genome U34 Set rc_AA924542_s_at - -
          Rat Genome U34 Set U73142_g_at - -
          Rat Genome U34 Set U73142_g_at - -
          Rat Genome U34 Set U73142_at - -
          Rat Genome U34 Set rc_AA924542_s_at - -
          Rat Genome U34 Set rc_AI137862_s_at - -
          Rat Genome U34 Set U91847_s_at - -
          Rat Genome U34 Set rc_AI171630_s_at - -
          Rat Genome U34 Set U73142_at - -
          Rat Genome U34 Set U91847_s_at - -
          Rat Genome U34 Set rc_AA924542_s_at - -
          Rat Genome U34 Set rc_AI137862_s_at - -
          Rat Genome U34 Set rc_AI171630_s_at - -
          Rat Genome U34 Set U73142_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21279379 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352809 - -
          RatRef-12 Expression BeadChip ILMN_48766 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline