Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578849_m1
          See other CSPG4 GE Assays ›
          Gene Symbol:
          Cspg4
          Gene Name:
          chondroitin sulfate proteoglycan 4
          Gene Aliases:
          Ng2
          Chromosome Location:
          Chr.8: 61532465 - 61567510 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578849_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cspg4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031022.1 NP_112284.1 1 - 2 159 135
          GenBank mRNA X56541.2 - 1 - 2 159 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000772.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cspg4

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          angiogenesis
          inflammatory response
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell proliferation
          glial cell migration
          response to lithium ion
          negative regulation of neuron projection development
          neuron remodeling
          intracellular signal transduction
          tissue remodeling
          positive regulation of peptidyl-tyrosine phosphorylation
          signal transducer activity
          collagen binding
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P696084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126023 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167557 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P696084 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P696084 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P696084 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P696084 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4642 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19002 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10910242 - -
          Affymetrix Rat Gene 1.0 ST Array 10910241 - -
          Affymetrix Rat Gene 1.0 ST Array 10910238 - -
          Affymetrix Rat Gene 1.0 ST Array 10910243 - -
          Affymetrix Rat Gene 1.0 ST Array 10910240 - -
          Affymetrix Rat Gene 1.0 ST Array 10910239 - -
          Affymetrix Rat Gene 1.0 ST Array 10910244 - -
          Affymetrix Rat Gene 1.0 ST Array 10910248 - -
          Affymetrix Rat Gene 1.0 ST Array 10910247 - -
          Affymetrix Rat Gene 1.0 ST Array 10910246 - -
          Affymetrix Rat Gene 1.0 ST Array 10910245 - -
          Affymetrix Rat Gene 1.1 ST Array 10910237 - -
          Rat Genome 230 2.0 Array 1368685_at - -
          Rat Genome 230 2.0 Array 1368685_PM_at - -
          Affymetrix HT Rat Focus Array 1368685_PM_at - -
          Affymetrix HT Rat Focus Array 1368685_at - -
          Affymetrix Rat Clariom S Assay PSR0800005197.rn.1 TC0800000772.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800005198.rn.1 TC0800000772.rn.1 RTA TC
          Rat Expression Set 230 1368685_at - -
          Rat Expression Set 230 1368685_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10910237 - -
          Rat Genome U34 Set X56541_at - -
          Rat Genome U34 Set rc_AA999096_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21517936 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352096 - -
          RatRef-12 Expression BeadChip ILMN_54579 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline