Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579020_m1
          See other HNMT GE Assays ›
          Gene Symbol:
          Hnmt
          Gene Name:
          histamine N-methyltransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 905111 - 937038 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579020_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnmt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031044.3 NP_112306.1 5 - 6 537 98
          GenBank mRNA BC087635.1 - 5 - 6 728 7319794 98
          D10693.1 - 5 - 6 537 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002629.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hnmt

          Gene Ontology Categories:

          Function(s) Process(es)

          response to tumor cell
          hyperosmotic response
          brain development
          response to amine
          methylation
          response to immobilization stress
          response to cocaine
          response to glucocorticoid
          response to interleukin-1
          N-methyltransferase activity
          histamine N-methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P389916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486801 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76739 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69961 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P389916 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P389916 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P389916 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P389916 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700769323 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701797389 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701829553 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33499 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48517 - -
          Agilent Rat cDNA Microarray (G4105A) 701829553 - -
          Agilent Rat cDNA Microarray (G4105A) 700769323 - -
          Agilent Rat cDNA Microarray (G4105A) 701797389 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843186 - -
          Affymetrix Rat Gene 1.0 ST Array 10843191 - -
          Affymetrix Rat Gene 1.0 ST Array 10843185 - -
          Affymetrix Rat Gene 1.0 ST Array 10843187 - -
          Affymetrix Rat Gene 1.0 ST Array 10843190 - -
          Affymetrix Rat Gene 1.0 ST Array 10843189 - -
          Affymetrix Rat Gene 1.0 ST Array 10843192 - -
          Affymetrix Rat Gene 1.0 ST Array 10843188 - -
          Affymetrix Rat Gene 1.1 ST Array 10843184 - -
          Rat Genome 230 2.0 Array 1385762_PM_at - -
          Rat Genome 230 2.0 Array 1387382_at - -
          Rat Genome 230 2.0 Array 1385762_at - -
          Rat Genome 230 2.0 Array 1387382_PM_at - -
          Affymetrix HT Rat Focus Array 1387382_at - -
          Affymetrix HT Rat Focus Array 1385762_PM_at - -
          Affymetrix HT Rat Focus Array 1387382_PM_at - -
          Affymetrix HT Rat Focus Array 1385762_at - -
          Affymetrix Rat Clariom S Assay PSR0300019472.rn.1 TC0300002629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019473.rn.1 TC0300002629.rn.1 RTA TC
          Rat Expression Set 230 1385762_at - -
          Rat Expression Set 230 1387382_PM_at - -
          Rat Expression Set 230 1385762_PM_at - -
          Rat Expression Set 230 1387382_at - -
          Affymetrix Rat Gene 1.0 ST Array 10843184 - -
          Rat Genome U34 Set D10693_at - -
          Rat Genome U34 Set S82579_s_at - -
          Rat Genome U34 Set rc_AI145151_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21331503 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363411 - -
          RatRef-12 Expression BeadChip ILMN_50405 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline