Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579159_m1
          See other MLH1 GE Assays ›
          Gene Symbol:
          Mlh1
          Gene Name:
          mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 119486655 - 119523716 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579159_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mlh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031053.1 NP_112315.1 9 - 10 790 65
          XM_008766652.1 XP_008764874.1 7 - 8 552 65
          GenBank mRNA U80054.1 - 9 - 10 790 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003440.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mlh1

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA poly(A) tail shortening
          resolution of meiotic recombination intermediates
          response to hypoxia
          somatic recombination of immunoglobulin genes involved in immune response
          DNA repair
          mismatch repair
          double-strand break repair via nonhomologous end joining
          cellular response to DNA damage stimulus
          male meiosis chromosome segregation
          meiotic nuclear division
          synapsis
          reciprocal meiotic recombination
          male meiosis
          spermatogenesis
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to toxic substance
          female meiosis chromosome segregation
          somatic hypermutation of immunoglobulin genes
          somatic recombination of immunoglobulin gene segments
          response to drug
          meiotic metaphase I plate congression
          meiotic chromosome segregation
          homologous chromosome segregation
          isotype switching
          negative regulation of mitotic recombination
          oogenesis
          meiotic spindle midzone assembly
          cellular response to organic cyclic compound
          molecular_function
          single-stranded DNA binding
          ATP binding
          ATPase activity
          mismatched DNA binding
          guanine/thymine mispair binding
          MutSalpha complex binding
          structure-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P142829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341150 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171881 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110893 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164232 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P142829 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P142829 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P142829 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P142829 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700415670 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102669 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27861 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83245 - -
          Agilent Rat cDNA Microarray (G4105A) 700415670 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10920576 - -
          Affymetrix Rat Gene 1.0 ST Array 10920584 - -
          Affymetrix Rat Gene 1.0 ST Array 10920582 - -
          Affymetrix Rat Gene 1.0 ST Array 10920575 - -
          Affymetrix Rat Gene 1.0 ST Array 10920580 - -
          Affymetrix Rat Gene 1.0 ST Array 10920574 - -
          Affymetrix Rat Gene 1.0 ST Array 10920581 - -
          Affymetrix Rat Gene 1.0 ST Array 10920570 - -
          Affymetrix Rat Gene 1.0 ST Array 10920583 - -
          Affymetrix Rat Gene 1.0 ST Array 10920578 - -
          Affymetrix Rat Gene 1.0 ST Array 10920585 - -
          Affymetrix Rat Gene 1.0 ST Array 10920579 - -
          Affymetrix Rat Gene 1.0 ST Array 10920569 - -
          Affymetrix Rat Gene 1.0 ST Array 10920571 - -
          Affymetrix Rat Gene 1.0 ST Array 10920587 - -
          Affymetrix Rat Gene 1.0 ST Array 10920586 - -
          Affymetrix Rat Gene 1.0 ST Array 10920577 - -
          Affymetrix Rat Gene 1.0 ST Array 10920573 - -
          Affymetrix Rat Gene 1.1 ST Array 10920568 - -
          Rat Genome 230 2.0 Array 1369965_at - -
          Rat Genome 230 2.0 Array 1369965_PM_at - -
          Affymetrix HT Rat Focus Array 1369965_at - -
          Affymetrix HT Rat Focus Array 1369965_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800028071.rn.1 TC0800003440.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800028072.rn.1 TC0800003440.rn.1 RTA TC
          Rat Expression Set 230 1369965_PM_at - -
          Rat Expression Set 230 1369965_at - -
          Affymetrix Rat Gene 1.0 ST Array 10920568 - -
          Rat Genome U34 Set U80054_at - -
          Rat Genome U34 Set U80054_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20976380 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline