Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579182_m1
          See other ALDH6A1 GE Assays ›
          Gene Symbol:
          Aldh6a1
          Gene Name:
          aldehyde dehydrogenase 6 family, member A1
          Gene Aliases:
          Mmsdh
          Chromosome Location:
          Chr.6: 108146552 - 108167185 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579182_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh6a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031057.2 NP_112319.2 11 - 12 1545 129
          XM_006240354.2 XP_006240416.1 11 - 12 1910 129
          XM_008764862.1 XP_008763084.1 10 - 11 1449 129
          XM_008764863.1 XP_008763085.1 11 - 12 1547 129
          GenBank mRNA CK476335.1 - 1 - 2 112 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002857.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aldh6a1

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          thymine catabolic process
          valine metabolic process
          valine catabolic process
          beta-alanine catabolic process
          thymine metabolic process
          brown fat cell differentiation
          oxidation-reduction process
          fatty-acyl-CoA binding
          aldehyde dehydrogenase (NAD) activity
          methylmalonate-semialdehyde dehydrogenase (acylating) activity
          thiolester hydrolase activity
          malonate-semialdehyde dehydrogenase (acetylating) activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P555261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953150 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124470 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P555261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P555261 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P555261 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2046 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48659 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75441 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891131 - -
          Affymetrix Rat Gene 1.0 ST Array 10891127 - -
          Affymetrix Rat Gene 1.0 ST Array 10891123 - -
          Affymetrix Rat Gene 1.0 ST Array 10891129 - -
          Affymetrix Rat Gene 1.0 ST Array 10891132 - -
          Affymetrix Rat Gene 1.0 ST Array 10891124 - -
          Affymetrix Rat Gene 1.0 ST Array 10891128 - -
          Affymetrix Rat Gene 1.0 ST Array 10891125 - -
          Affymetrix Rat Gene 1.0 ST Array 10891122 - -
          Affymetrix Rat Gene 1.0 ST Array 10891130 - -
          Affymetrix Rat Gene 1.0 ST Array 10891126 - -
          Affymetrix Rat Gene 1.0 ST Array 10891121 - -
          Affymetrix Rat Gene 1.1 ST Array 10891120 - -
          Rat Genome 230 2.0 Array 1387344_PM_at - -
          Rat Genome 230 2.0 Array 1368814_PM_at - -
          Rat Genome 230 2.0 Array 1368814_at - -
          Rat Genome 230 2.0 Array 1387344_at - -
          Affymetrix HT Rat Focus Array 1368814_at - -
          Affymetrix HT Rat Focus Array 1387344_at - -
          Affymetrix HT Rat Focus Array 1368814_PM_at - -
          Affymetrix HT Rat Focus Array 1387344_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600020137.rn.1 TC0600002857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600020136.rn.1 TC0600002857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600020135.rn.1 TC0600002857.rn.1 RTA TC
          Rat Expression Set 230 1368814_PM_at - -
          Rat Expression Set 230 1387344_PM_at - -
          Rat Expression Set 230 1368814_at - -
          Rat Expression Set 230 1387344_at - -
          Affymetrix Rat Gene 1.0 ST Array 10891120 - -
          Rat Genome U34 Set M93401_at - -
          Rat Genome U34 Set rc_AI171803_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20708753 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354510 - -
          RatRef-12 Expression BeadChip ILMN_53446 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline