Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579248_m1
          See other FEZ1 GE Assays ›
          Gene Symbol:
          Fez1
          Gene Name:
          fasciculation and elongation protein zeta 1 (zygin I)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 39305128 - 39350270 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579248_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fez1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031066.1 NP_112328.1 9 - 10 1321 66
          GenBank mRNA BC087740.1 - 9 - 10 1321 7314065 66
          FQ212529.1 - 7 - 8 1067 66
          U48249.2 - 9 - 10 1284 66
          U63740.1 - 9 - 10 1321 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000396.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fez1

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          axon guidance
          positive regulation of neuron projection development
          positive regulation of neuron differentiation
          establishment of mitochondrion localization
          cellular response to growth factor stimulus
          protein kinase C binding
          protein binding
          gamma-tubulin binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P334953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631609 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72716 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22873 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P334953 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P334953 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P334953 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P334953 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702154488 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701288808 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102924 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51939 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50727 - -
          Agilent Rat cDNA Microarray (G4105A) 701288808 - -
          Agilent Rat cDNA Microarray (G4105A) 702154488 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909028 - -
          Affymetrix Rat Gene 1.0 ST Array 10909029 - -
          Affymetrix Rat Gene 1.0 ST Array 10909023 - -
          Affymetrix Rat Gene 1.0 ST Array 10909030 - -
          Affymetrix Rat Gene 1.0 ST Array 10909026 - -
          Affymetrix Rat Gene 1.0 ST Array 10909031 - -
          Affymetrix Rat Gene 1.0 ST Array 10909024 - -
          Affymetrix Rat Gene 1.0 ST Array 10909032 - -
          Affymetrix Rat Gene 1.0 ST Array 10909027 - -
          Affymetrix Rat Gene 1.0 ST Array 10909025 - -
          Affymetrix Rat Gene 1.1 ST Array 10909022 - -
          Rat Genome 230 2.0 Array 1367918_at - -
          Rat Genome 230 2.0 Array 1367918_PM_at - -
          Affymetrix HT Rat Focus Array 1367918_at - -
          Affymetrix HT Rat Focus Array 1367918_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800002635.rn.1 TC0800000396.rn.1 RTA TC
          Rat Expression Set 230 1367918_at - -
          Rat Expression Set 230 1367918_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10909022 - -
          Rat Genome U34 Set U63740_at - -
          Rat Genome U34 Set U63740_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22406195 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355043 - -
          RatRef-12 Expression BeadChip ILMN_68428 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline