Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579419_m1
          See other PTGER2 GE Assays ›
          Gene Symbol:
          Ptger2
          Gene Name:
          prostaglandin E receptor 2 (subtype EP2)
          Gene Aliases:
          EP2, Ptger-ep2
          Chromosome Location:
          Chr.15: 19336039 - 19349759 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579419_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptger2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031088.1 NP_112350.1 1 - 2 883 71
          GenBank mRNA U48858.1 - 1 - 2 949 71
          U94708.1 - 1 - 2 883 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000191.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptger2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          response to lipopolysaccharide
          regulation of cell proliferation
          cellular response to prostaglandin E stimulus
          prostaglandin E receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12508 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12508 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12508 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12508 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12508 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10779554 - -
          Affymetrix Rat Gene 1.0 ST Array 10779585 - -
          Affymetrix Rat Gene 1.0 ST Array 10779588 - -
          Affymetrix Rat Gene 1.0 ST Array 10779555 - -
          Affymetrix Rat Gene 1.0 ST Array 10779557 - -
          Affymetrix Rat Gene 1.0 ST Array 10779587 - -
          Affymetrix Rat Gene 1.0 ST Array 10779586 - -
          Affymetrix Rat Gene 1.0 ST Array 10779556 - -
          Affymetrix Rat Gene 1.1 ST Array 10779584 - -
          Affymetrix Rat Gene 1.1 ST Array 10779553 - -
          Rat Genome 230 2.0 Array 1369766_PM_at - -
          Rat Genome 230 2.0 Array 1385434_at - -
          Rat Genome 230 2.0 Array 1369766_at - -
          Rat Genome 230 2.0 Array 1385434_PM_at - -
          Affymetrix HT Rat Focus Array 1385434_PM_at - -
          Affymetrix HT Rat Focus Array 1385434_at - -
          Affymetrix HT Rat Focus Array 1369766_PM_at - -
          Affymetrix HT Rat Focus Array 1369766_at - -
          Affymetrix Rat Clariom S Assay PSR1500001420.rn.1 TC1500000191.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001418.rn.1 TC1500000191.rn.1 RTA TC
          Rat Expression Set 230 1369766_at - -
          Rat Expression Set 230 1369766_PM_at - -
          Rat Expression Set 230 1385434_at - -
          Rat Expression Set 230 1385434_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10779584 - -
          Affymetrix Rat Gene 1.0 ST Array 10779553 - -
          Rat Genome U34 Set U94708_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21949308 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361292 - -
          RatRef-12 Expression BeadChip ILMN_62420 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline