Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579490_m1
          See other ROCK1 GE Assays ›
          Gene Symbol:
          Rock1
          Gene Name:
          Rho-associated coiled-coil containing protein kinase 1
          Gene Aliases:
          P160Rock, ROCK-I
          Chromosome Location:
          Chr.18: 1273585 - 1390790 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579490_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rock1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031098.1 NP_112360.1 16 - 17 1994 75
          XM_006254400.2 XP_006254462.1 16 - 17 2746 75
          XM_006254401.2 XP_006254463.1 16 - 17 2743 75
          XM_008771957.1 XP_008770179.1 16 - 17 2746 75
          GenBank mRNA AY325220.1 - 461 75
          FQ225745.1 - 17 - 18 2470 75
          U61266.1 - 16 - 17 1994 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001011.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rock1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          apical constriction
          protein phosphorylation
          apoptotic process
          cytoskeleton organization
          leukocyte cell-cell adhesion
          Rho protein signal transduction
          negative regulation of angiogenesis
          membrane to membrane docking
          actin cytoskeleton organization
          bleb assembly
          regulation of actin filament-based process
          negative regulation of neuron apoptotic process
          regulation of keratinocyte differentiation
          leukocyte migration
          leukocyte tethering or rolling
          myoblast migration
          regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          regulation of establishment of cell polarity
          protein serine/threonine kinase activity
          ATP binding
          GTP-Rho binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144377 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46221 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55005 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94950 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018164 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018164 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018164 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039856 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700522707 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702143230 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45631 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100368 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48863 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67065 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32987 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57027 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74378 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40361 - -
          Agilent Rat cDNA Microarray (G4105A) 700522707 - -
          Agilent Rat cDNA Microarray (G4105A) 702143230 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803173 - -
          Affymetrix Rat Gene 1.0 ST Array 10803179 - -
          Affymetrix Rat Gene 1.0 ST Array 10803186 - -
          Affymetrix Rat Gene 1.0 ST Array 10803162 - -
          Affymetrix Rat Gene 1.0 ST Array 10803191 - -
          Affymetrix Rat Gene 1.0 ST Array 10803167 - -
          Affymetrix Rat Gene 1.0 ST Array 10803183 - -
          Affymetrix Rat Gene 1.0 ST Array 10803181 - -
          Affymetrix Rat Gene 1.0 ST Array 10803177 - -
          Affymetrix Rat Gene 1.0 ST Array 10803180 - -
          Affymetrix Rat Gene 1.0 ST Array 10803174 - -
          Affymetrix Rat Gene 1.0 ST Array 10803161 - -
          Affymetrix Rat Gene 1.0 ST Array 10803176 - -
          Affymetrix Rat Gene 1.0 ST Array 10803170 - -
          Affymetrix Rat Gene 1.0 ST Array 10803189 - -
          Affymetrix Rat Gene 1.0 ST Array 10803178 - -
          Affymetrix Rat Gene 1.0 ST Array 10803166 - -
          Affymetrix Rat Gene 1.0 ST Array 10803171 - -
          Affymetrix Rat Gene 1.0 ST Array 10803160 - -
          Affymetrix Rat Gene 1.0 ST Array 10803175 - -
          Affymetrix Rat Gene 1.0 ST Array 10803190 - -
          Affymetrix Rat Gene 1.0 ST Array 10803188 - -
          Affymetrix Rat Gene 1.0 ST Array 10803187 - -
          Affymetrix Rat Gene 1.0 ST Array 10803184 - -
          Affymetrix Rat Gene 1.0 ST Array 10803163 - -
          Affymetrix Rat Gene 1.0 ST Array 10803172 - -
          Affymetrix Rat Gene 1.0 ST Array 10803165 - -
          Affymetrix Rat Gene 1.0 ST Array 10803164 - -
          Affymetrix Rat Gene 1.0 ST Array 10803185 - -
          Affymetrix Rat Gene 1.0 ST Array 10803182 - -
          Affymetrix Rat Gene 1.0 ST Array 10803169 - -
          Affymetrix Rat Gene 1.1 ST Array 10803158 - -
          Rat Genome 230 2.0 Array 1368932_PM_at - -
          Rat Genome 230 2.0 Array 1368932_at - -
          Affymetrix HT Rat Focus Array 1368932_at - -
          Affymetrix HT Rat Focus Array 1368932_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800007198.rn.1 TC1800001011.rn.1 RTA TC
          Rat Expression Set 230 1368932_at - -
          Rat Expression Set 230 1368932_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803158 - -
          Rat Genome U34 Set rc_AI137331_at - -
          Rat Genome U34 Set U61266_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21682418 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366295 - -
          RatRef-12 Expression BeadChip ILMN_50836 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline