Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579605_m1
          See other SOAT1 GE Assays ›
          Gene Symbol:
          Soat1
          Gene Name:
          sterol O-acyltransferase 1
          Gene Aliases:
          Acat-1
          Chromosome Location:
          Chr.13: 74032622 - 74077759 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579605_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Soat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031118.1 NP_112380.1 4 - 5 403 76
          XM_008769681.1 XP_008767903.1 3 - 4 652 76
          GenBank mRNA D86373.1 - 4 - 5 403 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001694.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Rat Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Soat1

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          cholesterol metabolic process
          macrophage derived foam cell differentiation
          cholesterol storage
          cholesterol efflux
          very-low-density lipoprotein particle assembly
          cholesterol esterification
          positive regulation of amyloid precursor protein biosynthetic process
          fatty-acyl-CoA binding
          sterol O-acyltransferase activity
          cholesterol binding
          cholesterol O-acyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P170652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335032 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72287 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124814 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140936 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P170652 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P170652 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P170652 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P170652 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97978 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43555 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10768984 - -
          Affymetrix Rat Gene 1.0 ST Array 10768990 - -
          Affymetrix Rat Gene 1.0 ST Array 10768995 - -
          Affymetrix Rat Gene 1.0 ST Array 10768991 - -
          Affymetrix Rat Gene 1.0 ST Array 10768989 - -
          Affymetrix Rat Gene 1.0 ST Array 10768986 - -
          Affymetrix Rat Gene 1.0 ST Array 10768992 - -
          Affymetrix Rat Gene 1.0 ST Array 10768988 - -
          Affymetrix Rat Gene 1.0 ST Array 10768987 - -
          Affymetrix Rat Gene 1.0 ST Array 10768981 - -
          Affymetrix Rat Gene 1.0 ST Array 10768980 - -
          Affymetrix Rat Gene 1.0 ST Array 10768982 - -
          Affymetrix Rat Gene 1.0 ST Array 10768993 - -
          Affymetrix Rat Gene 1.0 ST Array 10768994 - -
          Affymetrix Rat Gene 1.0 ST Array 10768985 - -
          Affymetrix Rat Gene 1.0 ST Array 10768983 - -
          Affymetrix Rat Gene 1.1 ST Array 10768979 - -
          Rat Genome 230 2.0 Array 1379835_PM_at - -
          Rat Genome 230 2.0 Array 1373869_at - -
          Rat Genome 230 2.0 Array 1373869_PM_at - -
          Rat Genome 230 2.0 Array 1369002_at - -
          Rat Genome 230 2.0 Array 1379835_at - -
          Rat Genome 230 2.0 Array 1369002_PM_at - -
          Rat Genome 230 2.0 Array 1374976_a_at - -
          Rat Genome 230 2.0 Array 1374976_PM_a_at - -
          Affymetrix HT Rat Focus Array 1373869_at - -
          Affymetrix HT Rat Focus Array 1374976_a_at - -
          Affymetrix HT Rat Focus Array 1373869_PM_at - -
          Affymetrix HT Rat Focus Array 1369002_at - -
          Affymetrix HT Rat Focus Array 1374976_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369002_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300011844.rn.1 TC1300001694.rn.1 RTA TC
          Rat Expression Set 230 1373869_PM_at - -
          Rat Expression Set 230 1374976_PM_a_at - -
          Rat Expression Set 230 1374976_a_at - -
          Rat Expression Set 230 1369002_at - -
          Rat Expression Set 230 1373869_at - -
          Rat Expression Set 230 1369002_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10768979 - -
          Rat Genome U34 Set D86373_s_at - -
          Rat Genome U34 Set rc_AI236053_at - -
          Rat Genome U34 Set rc_AI236009_at - -
          Rat Genome U34 Set rc_AI103060_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21423884 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350250 - -
          RatRef-12 Expression BeadChip ILMN_52563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline