Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579648_m1
          See other STX4 GE Assays ›
          Gene Symbol:
          Stx4
          Gene Name:
          syntaxin 4
          Gene Aliases:
          Stx4a
          Chromosome Location:
          Chr.1: 199287386 - 199295609 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579648_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stx4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031125.1 NP_112387.1 5 - 6 405 121
          XM_006230345.2 XP_006230407.1 5 - 6 912 121
          XM_006230346.2 XP_006230408.1 5 - 6 937 121
          XM_006230348.2 XP_006230410.1 5 - 6 902 121
          GenBank mRNA L20821.1 - 5 - 6 405 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002664.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stx4

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          intracellular protein transport
          positive regulation of cell proliferation
          synaptic vesicle docking
          regulation of exocytosis
          positive regulation of cell migration
          synaptic vesicle fusion to presynaptic active zone membrane
          response to hydroperoxide
          SNARE complex assembly
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          positive regulation of catalytic activity
          positive regulation of eosinophil degranulation
          positive regulation of cell adhesion
          intracellular transport
          organelle fusion
          positive regulation of chemotaxis
          positive regulation of immunoglobulin secretion
          long-term synaptic potentiation
          membrane fusion
          positive regulation of establishment of protein localization to plasma membrane
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of protein localization to plasma membrane
          positive regulation of protein localization to cell surface
          SNARE binding
          SNAP receptor activity
          protein binding
          sphingomyelin phosphodiesterase activator activity
          Rab GTPase binding
          spectrin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P420131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135442 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P420131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P420131 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P420131 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P420131 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700369894 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70025 - -
          Agilent Rat cDNA Microarray (G4105A) 700369894 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10711226 - -
          Affymetrix Rat Gene 1.0 ST Array 10711224 - -
          Affymetrix Rat Gene 1.0 ST Array 10711225 - -
          Affymetrix Rat Gene 1.0 ST Array 10711220 - -
          Affymetrix Rat Gene 1.0 ST Array 10711228 - -
          Affymetrix Rat Gene 1.0 ST Array 10711223 - -
          Affymetrix Rat Gene 1.0 ST Array 10711230 - -
          Affymetrix Rat Gene 1.0 ST Array 10711221 - -
          Affymetrix Rat Gene 1.0 ST Array 10711229 - -
          Affymetrix Rat Gene 1.0 ST Array 10711222 - -
          Affymetrix Rat Gene 1.0 ST Array 10711227 - -
          Affymetrix Rat Gene 1.1 ST Array 10711219 - -
          Rat Genome 230 2.0 Array 1370014_PM_at - -
          Rat Genome 230 2.0 Array 1370014_at - -
          Affymetrix HT Rat Focus Array 1370014_at - -
          Affymetrix HT Rat Focus Array 1370014_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100020707.rn.1 TC0100002664.rn.1 RTA TC
          Rat Expression Set 230 1370014_at - -
          Rat Expression Set 230 1370014_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10711219 - -
          Rat Genome U34 Set L20821_at - -
          Rat Genome U34 Set rc_AI011416_at - -
          Rat Genome U34 Set rc_AA850008_s_at - -
          Rat Genome U34 Set L20821_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21288630 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373557 - -
          RatRef-12 Expression BeadChip ILMN_51471 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline