Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579697_m1
          See other KLF10 GE Assays ›
          Gene Symbol:
          Klf10
          Gene Name:
          Kruppel-like factor 10
          Gene Aliases:
          Tieg
          Chromosome Location:
          Chr.7: 77156010 - 77162096 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579697_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klf10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031135.2 NP_112397.1 1 - 2 84 78
          GenBank mRNA BC097309.1 - 1 - 2 84 7387595 78
          U78875.1 - 1 - 2 353 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003028.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klf10

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          circadian rhythm
          cellular response to starvation
          bone mineralization
          somatic stem cell population maintenance
          regulation of circadian rhythm
          positive regulation of osteoclast differentiation
          negative regulation of transcription, DNA-templated
          cellular response to peptide
          core promoter sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776268 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12535 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12535 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12535 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12535 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P526729 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700494824 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700634678 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701509747 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79095 - -
          Agilent Rat cDNA Microarray (G4105A) 701509747 - -
          Agilent Rat cDNA Microarray (G4105A) 700634678 - -
          Agilent Rat cDNA Microarray (G4105A) 700494824 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903460 - -
          Affymetrix Rat Gene 1.0 ST Array 10903465 - -
          Affymetrix Rat Gene 1.0 ST Array 10903462 - -
          Affymetrix Rat Gene 1.0 ST Array 10903464 - -
          Affymetrix Rat Gene 1.0 ST Array 10903461 - -
          Affymetrix Rat Gene 1.0 ST Array 10903463 - -
          Affymetrix Rat Gene 1.1 ST Array 10903459 - -
          Rat Genome 230 2.0 Array 1368650_at - -
          Rat Genome 230 2.0 Array 1368650_PM_at - -
          Affymetrix HT Rat Focus Array 1368650_PM_at - -
          Affymetrix HT Rat Focus Array 1368650_at - -
          Affymetrix Rat Clariom S Assay PSR0700023481.rn.1 TC0700003028.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023479.rn.1 TC0700003028.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023476.rn.1 TC0700003028.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023478.rn.1 TC0700003028.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023480.rn.1 TC0700003028.rn.1 RTA TC
          Rat Expression Set 230 1368650_PM_at - -
          Rat Expression Set 230 1368650_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903459 - -
          Rat Genome U34 Set rc_AI172476_at - -
          Rat Genome U34 Set U88630_at - -
          Rat Genome U34 Set rc_AI071299_at - -
          Rat Genome U34 Set U88630_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21305156 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362189 - -
          RatRef-12 Expression BeadChip ILMN_61759 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline