Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579942_m1
          See other CDS1 GE Assays ›
          Gene Symbol:
          Cds1
          Gene Name:
          CDP-diacylglycerol synthase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 9396511 - 9456964 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579942_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cds1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031242.2 NP_112521.2 1 - 2 176 69
          GenBank mRNA AB009999.1 - 121 69
          BC127492.1 - 1 - 2 176 8373547 69
          FQ212493.1 - 151 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001365.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cds1

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylinositol biosynthetic process
          CDP-diacylglycerol biosynthetic process
          phosphatidate cytidylyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P320073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824813 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67875 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99857 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P320073 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P320073 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P320073 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P320073 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P598531 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P294932 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700887369 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700370295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85952 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98498 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75519 - -
          Agilent Rat cDNA Microarray (G4105A) 700370295 - -
          Agilent Rat cDNA Microarray (G4105A) 700887369 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775504 - -
          Affymetrix Rat Gene 1.0 ST Array 10775513 - -
          Affymetrix Rat Gene 1.0 ST Array 10775515 - -
          Affymetrix Rat Gene 1.0 ST Array 10775508 - -
          Affymetrix Rat Gene 1.0 ST Array 10775518 - -
          Affymetrix Rat Gene 1.0 ST Array 10775505 - -
          Affymetrix Rat Gene 1.0 ST Array 10775506 - -
          Affymetrix Rat Gene 1.0 ST Array 10775507 - -
          Affymetrix Rat Gene 1.0 ST Array 10775512 - -
          Affymetrix Rat Gene 1.0 ST Array 10775514 - -
          Affymetrix Rat Gene 1.0 ST Array 10775509 - -
          Affymetrix Rat Gene 1.0 ST Array 10775511 - -
          Affymetrix Rat Gene 1.0 ST Array 10775516 - -
          Affymetrix Rat Gene 1.0 ST Array 10775517 - -
          Affymetrix Rat Gene 1.0 ST Array 10775510 - -
          Affymetrix Rat Gene 1.1 ST Array 10775503 - -
          Rat Genome 230 2.0 Array 1368248_at - -
          Rat Genome 230 2.0 Array 1368248_PM_at - -
          Affymetrix HT Rat Focus Array 1368248_at - -
          Affymetrix HT Rat Focus Array 1368248_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400010192.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010197.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010198.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010187.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010195.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010186.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010188.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010191.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010190.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010193.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010201.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010202.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010196.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010200.rn.1 TC1400001365.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010199.rn.1 TC1400001365.rn.1 RTA TC
          Rat Expression Set 230 1368248_at - -
          Rat Expression Set 230 1368248_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775503 - -
          Rat Genome U34 Set AB009999_g_at - -
          Rat Genome U34 Set AB009999_at - -
          Rat Genome U34 Set AB000517_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20836072 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352002 - -
          RatRef-12 Expression BeadChip ILMN_56455 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline