Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580039_m1
          See other PREP GE Assays ›
          Gene Symbol:
          Prep
          Gene Name:
          prolyl endopeptidase
          Gene Aliases:
          rPop
          Chromosome Location:
          Chr.20: 50172618 - 50269992 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580039_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prep
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031324.1 NP_112614.1 9 - 10 1349 90
          GenBank mRNA AB012759.1 - 9 - 10 1349 90
          FQ213388.1 - 7 - 8 885 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000886.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prep

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          endopeptidase activity
          serine-type endopeptidase activity
          peptide binding
          serine-type exopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171080 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163789 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99953 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28778 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21637 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60533 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14875 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14875 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14875 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14875 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55144 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31262 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57697 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7572 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8245 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47764 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101406 - -
          Agilent Rat cDNA Microarray (G4105A) 701285040 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830656 - -
          Affymetrix Rat Gene 1.0 ST Array 10830665 - -
          Affymetrix Rat Gene 1.0 ST Array 10830661 - -
          Affymetrix Rat Gene 1.0 ST Array 10830660 - -
          Affymetrix Rat Gene 1.0 ST Array 10830667 - -
          Affymetrix Rat Gene 1.0 ST Array 10830668 - -
          Affymetrix Rat Gene 1.0 ST Array 10830666 - -
          Affymetrix Rat Gene 1.0 ST Array 10830655 - -
          Affymetrix Rat Gene 1.0 ST Array 10830658 - -
          Affymetrix Rat Gene 1.0 ST Array 10830659 - -
          Affymetrix Rat Gene 1.0 ST Array 10830662 - -
          Affymetrix Rat Gene 1.0 ST Array 10830657 - -
          Affymetrix Rat Gene 1.0 ST Array 10830669 - -
          Affymetrix Rat Gene 1.0 ST Array 10830664 - -
          Affymetrix Rat Gene 1.0 ST Array 10830670 - -
          Affymetrix Rat Gene 1.0 ST Array 10830663 - -
          Affymetrix Rat Gene 1.1 ST Array 10830654 - -
          Rat Genome 230 2.0 Array 1368234_at - -
          Rat Genome 230 2.0 Array 1390855_at - -
          Rat Genome 230 2.0 Array 1390855_PM_at - -
          Rat Genome 230 2.0 Array 1368234_PM_at - -
          Affymetrix HT Rat Focus Array 1368234_PM_at - -
          Affymetrix HT Rat Focus Array 1368234_at - -
          Affymetrix Rat Clariom S Assay PSR2000007363.rn.1 TC2000000886.rn.1 RTA TC
          Rat Expression Set 230 1368234_PM_at - -
          Rat Expression Set 230 1368234_at - -
          Affymetrix Rat Gene 1.0 ST Array 10830654 - -
          Rat Genome U34 Set AB012759_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22178080 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369198 - -
          RatRef-12 Expression BeadChip ILMN_49092 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline