Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580082_m1
          See other SLC22A8 GE Assays ›
          Gene Symbol:
          Slc22a8
          Gene Name:
          solute carrier family 22 (organic anion transporter), member 8
          Gene Aliases:
          OCT3, Oat3, Roct
          Chromosome Location:
          Chr.1: 224800080 - 224818482 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580082_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc22a8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031332.1 NP_112622.1 5 - 6 879 64
          XM_008760243.1 XP_008758465.1 4 - 5 694 64
          GenBank mRNA AB017446.1 - 5 - 6 879 64
          BC081777.1 - 5 - 6 877 7109603 64
          FQ209755.1 - 891 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003145.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc22a8

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          response to toxic substance
          quaternary ammonium group transport
          response to methotrexate
          glutathione transport
          anion transmembrane transport
          protein kinase C binding
          inorganic anion exchanger activity
          organic anion transmembrane transporter activity
          quaternary ammonium group transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82167 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052225 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052225 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P543342 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701430253 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701263974 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68168 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28227 - -
          Agilent Rat cDNA Microarray (G4105A) 701430253 - -
          Agilent Rat cDNA Microarray (G4105A) 701263974 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713557 - -
          Affymetrix Rat Gene 1.0 ST Array 10713559 - -
          Affymetrix Rat Gene 1.0 ST Array 10713558 - -
          Affymetrix Rat Gene 1.0 ST Array 10713561 - -
          Affymetrix Rat Gene 1.0 ST Array 10713567 - -
          Affymetrix Rat Gene 1.0 ST Array 10713564 - -
          Affymetrix Rat Gene 1.0 ST Array 10713563 - -
          Affymetrix Rat Gene 1.0 ST Array 10713565 - -
          Affymetrix Rat Gene 1.0 ST Array 10713560 - -
          Affymetrix Rat Gene 1.0 ST Array 10713556 - -
          Affymetrix Rat Gene 1.0 ST Array 10713562 - -
          Affymetrix Rat Gene 1.1 ST Array 10713555 - -
          Rat Genome 230 2.0 Array 1368461_PM_at - -
          Rat Genome 230 2.0 Array 1368461_at - -
          Affymetrix HT Rat Focus Array 1368461_PM_at - -
          Affymetrix HT Rat Focus Array 1368461_at - -
          Affymetrix Rat Clariom S Assay PSR0100025716.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025725.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025728.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025723.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025724.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025727.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025721.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025715.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025722.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025726.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025718.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025719.rn.1 TC0100003145.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025714.rn.1 TC0100003145.rn.1 RTA TC
          Rat Expression Set 230 1368461_at - -
          Rat Expression Set 230 1368461_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713555 - -
          Rat Genome U34 Set rc_AI169656_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20967193 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358346 - -
          RatRef-12 Expression BeadChip ILMN_68432 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline