Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580133_m1
          See other CAMKK2 GE Assays ›
          Gene Symbol:
          Camkk2
          Gene Name:
          calcium/calmodulin-dependent protein kinase kinase 2, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 39253409 - 39302601 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580133_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Camkk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031338.1 NP_112628.1 14 - 15 1575 73
          GenBank mRNA AB018081.1 - 14 - 15 1575 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000636.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Camkk2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          activation of protein kinase activity
          protein autophosphorylation
          protein kinase activity
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          calcium ion binding
          calmodulin binding
          ATP binding
          protein kinase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705438 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119022 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158296 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104268 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66220 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150331 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25804 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49105 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12559 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12559 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12559 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12559 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702033992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59896 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34241 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32767 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98586 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37647 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101769 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37058 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84486 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43489 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83047 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86242 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36706 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66405 - -
          Agilent Rat cDNA Microarray (G4105A) 702033992 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10758518 - -
          Affymetrix Rat Gene 1.0 ST Array 10758522 - -
          Affymetrix Rat Gene 1.0 ST Array 10758526 - -
          Affymetrix Rat Gene 1.0 ST Array 10758530 - -
          Affymetrix Rat Gene 1.0 ST Array 10758527 - -
          Affymetrix Rat Gene 1.0 ST Array 10758521 - -
          Affymetrix Rat Gene 1.0 ST Array 10758528 - -
          Affymetrix Rat Gene 1.0 ST Array 10758520 - -
          Affymetrix Rat Gene 1.0 ST Array 10758519 - -
          Affymetrix Rat Gene 1.0 ST Array 10758524 - -
          Affymetrix Rat Gene 1.0 ST Array 10758532 - -
          Affymetrix Rat Gene 1.0 ST Array 10758525 - -
          Affymetrix Rat Gene 1.0 ST Array 10758529 - -
          Affymetrix Rat Gene 1.0 ST Array 10758531 - -
          Affymetrix Rat Gene 1.0 ST Array 10758523 - -
          Affymetrix Rat Gene 1.0 ST Array 10758533 - -
          Affymetrix Rat Gene 1.1 ST Array 10758517 - -
          Rat Genome 230 2.0 Array 1375025_at - -
          Rat Genome 230 2.0 Array 1368753_at - -
          Rat Genome 230 2.0 Array 1375025_PM_at - -
          Rat Genome 230 2.0 Array 1368753_PM_at - -
          Affymetrix HT Rat Focus Array 1375025_PM_at - -
          Affymetrix HT Rat Focus Array 1368753_PM_at - -
          Affymetrix HT Rat Focus Array 1375025_at - -
          Affymetrix HT Rat Focus Array 1368753_at - -
          Affymetrix Rat Clariom S Assay PSR1200005617.rn.1 TC1200000636.rn.1 RTA TC
          Rat Expression Set 230 1375025_at - -
          Rat Expression Set 230 1375025_PM_at - -
          Rat Expression Set 230 1368753_at - -
          Rat Expression Set 230 1368753_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10758517 - -
          Rat Genome U34 Set AB018081_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21770302 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370123 - -
          RatRef-12 Expression BeadChip ILMN_51689 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline