Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580189_m1
          See other SLC7A7 GE Assays ›
          Gene Symbol:
          Slc7a7
          Gene Name:
          solute carrier family 7 (amino acid transporter light chain, y+L system), member 7
          Gene Aliases:
          y+LAT1
          Chromosome Location:
          Chr.15: 33013353 - 33059739 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580189_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc7a7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031341.1 NP_112631.1 4 - 5 975 91
          XM_006252020.2 XP_006252082.1 4 - 5 911 91
          XM_006252021.2 XP_006252083.1 7 - 8 1380 91
          XM_006252023.2 XP_006252085.1 4 - 5 916 91
          XM_006252024.2 XP_006252086.1 4 - 5 1605 91
          XM_008770705.1 XP_008768927.1 6 - 7 1323 91
          GenBank mRNA AB020520.1 - 4 - 5 975 91
          AF200684.1 - 3 - 4 783 91
          BC091142.1 - 4 - 5 890 7129657 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001668.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc7a7

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of arginine metabolic process
          amino acid transmembrane transport
          biological_process
          molecular_function
          amino acid transmembrane transporter activity
          basic amino acid transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P538658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116995 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79797 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119771 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538658 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538658 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P538658 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P538658 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701287466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39813 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17491 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9375 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42634 - -
          Agilent Rat cDNA Microarray (G4105A) 701287466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783547 - -
          Affymetrix Rat Gene 1.0 ST Array 10783538 - -
          Affymetrix Rat Gene 1.0 ST Array 10783543 - -
          Affymetrix Rat Gene 1.0 ST Array 10783541 - -
          Affymetrix Rat Gene 1.0 ST Array 10783539 - -
          Affymetrix Rat Gene 1.0 ST Array 10783540 - -
          Affymetrix Rat Gene 1.0 ST Array 10783549 - -
          Affymetrix Rat Gene 1.0 ST Array 10783546 - -
          Affymetrix Rat Gene 1.0 ST Array 10783545 - -
          Affymetrix Rat Gene 1.0 ST Array 10783548 - -
          Affymetrix Rat Gene 1.0 ST Array 10783542 - -
          Affymetrix Rat Gene 1.0 ST Array 10783544 - -
          Affymetrix Rat Gene 1.1 ST Array 10783537 - -
          Rat Genome 230 2.0 Array 1387808_at - -
          Rat Genome 230 2.0 Array 1387808_PM_at - -
          Affymetrix HT Rat Focus Array 1387808_PM_at - -
          Affymetrix HT Rat Focus Array 1387808_at - -
          Affymetrix Rat Clariom S Assay PSR1500010493.rn.1 TC1500001668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010489.rn.1 TC1500001668.rn.1 RTA TC
          Rat Expression Set 230 1387808_at - -
          Rat Expression Set 230 1387808_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783537 - -
          Rat Genome U34 Set rc_AI169077_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20879733 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358568 - -
          RatRef-12 Expression BeadChip ILMN_56785 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline