Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580232_m1
          See other PTBP3 GE Assays ›
          Gene Symbol:
          Ptbp3
          Gene Name:
          polypyrimidine tract binding protein 3
          Gene Aliases:
          Rod1
          Chromosome Location:
          Chr.5: 76670074 - 76756195 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580232_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptbp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031346.1 NP_112636.1 8 - 9 1067 64
          XM_006238206.2 XP_006238268.1 9 - 10 1155 64
          XM_006238207.2 XP_006238269.1 9 - 10 1147 64
          XM_006238208.2 XP_006238270.1 8 - 9 1112 64
          XM_006238209.2 XP_006238271.1 6 - 7 759 64
          GenBank mRNA AB023966.1 - 8 - 9 1067 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002851.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptbp3

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          negative regulation of RNA splicing
          erythrocyte maturation
          regulation of cell differentiation
          negative regulation of mRNA splicing, via spliceosome
          nucleotide binding
          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P471901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208791 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36452 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50325 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P471901 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P471901 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P471901 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P471901 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17070 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97838 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42282 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72342 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9619 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78772 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877192 - -
          Affymetrix Rat Gene 1.0 ST Array 10877194 - -
          Affymetrix Rat Gene 1.0 ST Array 10877205 - -
          Affymetrix Rat Gene 1.0 ST Array 10877195 - -
          Affymetrix Rat Gene 1.0 ST Array 10877191 - -
          Affymetrix Rat Gene 1.0 ST Array 10877196 - -
          Affymetrix Rat Gene 1.0 ST Array 10877200 - -
          Affymetrix Rat Gene 1.0 ST Array 10877201 - -
          Affymetrix Rat Gene 1.0 ST Array 10877197 - -
          Affymetrix Rat Gene 1.0 ST Array 10877202 - -
          Affymetrix Rat Gene 1.0 ST Array 10877198 - -
          Affymetrix Rat Gene 1.0 ST Array 10877190 - -
          Affymetrix Rat Gene 1.0 ST Array 10877193 - -
          Affymetrix Rat Gene 1.0 ST Array 10877199 - -
          Affymetrix Rat Gene 1.1 ST Array 10877189 - -
          Rat Genome 230 2.0 Array 1387432_PM_at - -
          Rat Genome 230 2.0 Array 1382005_PM_at - -
          Rat Genome 230 2.0 Array 1395574_at - -
          Rat Genome 230 2.0 Array 1382005_at - -
          Rat Genome 230 2.0 Array 1395574_PM_at - -
          Rat Genome 230 2.0 Array 1387432_at - -
          Affymetrix HT Rat Focus Array 1387432_PM_at - -
          Affymetrix HT Rat Focus Array 1387432_at - -
          Affymetrix Rat Clariom S Assay PSR0500021157.rn.1 TC0500002851.rn.1 RTA TC
          Rat Expression Set 230 1387432_PM_at - -
          Rat Expression Set 230 1387432_at - -
          Affymetrix Rat Gene 1.0 ST Array 10877189 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21196384 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352656 - -
          RatRef-12 Expression BeadChip ILMN_67721 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline