Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580247_m1
          See other FCNA GE Assays ›
          Gene Symbol:
          Fcna
          Gene Name:
          ficolin A
          Gene Aliases:
          Fcn1
          Chromosome Location:
          Chr.3: 2850006 - 2853230 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580247_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fcna
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031348.2 NP_112638.2 5 - 6 420 131
          XM_008761614.1 XP_008759836.1 5 - 6 443 131
          GenBank mRNA AB026057.2 - 5 - 6 331 131
          BC088128.1 - 5 - 6 420 7376326 131
          FQ227647.1 - 5 - 6 440 131
          FQ234790.1 - 5 - 6 440 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002694.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fcna

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          complement activation, lectin pathway
          cell surface pattern recognition receptor signaling pathway
          G-protein coupled receptor signaling pathway
          innate immune response
          positive regulation of interleukin-8 secretion
          signaling pattern recognition receptor activity
          carbohydrate binding
          metal ion binding
          carbohydrate derivative binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P780932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682309 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P780932 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P780932 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P780932 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P780932 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700543784 - -
          Agilent Rat cDNA Microarray (G4105A) 700543784 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843585 - -
          Affymetrix Rat Gene 1.0 ST Array 10843586 - -
          Affymetrix Rat Gene 1.0 ST Array 10843581 - -
          Affymetrix Rat Gene 1.0 ST Array 10843584 - -
          Affymetrix Rat Gene 1.0 ST Array 10843580 - -
          Affymetrix Rat Gene 1.0 ST Array 10843579 - -
          Affymetrix Rat Gene 1.0 ST Array 10843576 - -
          Affymetrix Rat Gene 1.0 ST Array 10843582 - -
          Affymetrix Rat Gene 1.0 ST Array 10843578 - -
          Affymetrix Rat Gene 1.0 ST Array 10843583 - -
          Affymetrix Rat Gene 1.0 ST Array 10843577 - -
          Affymetrix Rat Gene 1.0 ST Array 10843587 - -
          Affymetrix Rat Gene 1.1 ST Array 10843575 - -
          Rat Genome 230 2.0 Array 1387794_PM_at - -
          Rat Genome 230 2.0 Array 1387794_at - -
          Affymetrix HT Rat Focus Array 1387794_PM_at - -
          Affymetrix HT Rat Focus Array 1387794_at - -
          Affymetrix Rat Clariom S Assay PSR0300020202.rn.1 TC0300002694.rn.1 RTA TC
          Rat Expression Set 230 1387794_at - -
          Rat Expression Set 230 1387794_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10843575 - -
          Rat Genome U34 Set rc_AI104596_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21616800 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349211 - -
          RatRef-12 Expression BeadChip ILMN_64679 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline