Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580302_m1
          See other DBNL GE Assays ›
          Gene Symbol:
          Dbnl
          Gene Name:
          drebrin-like
          Gene Aliases:
          Sh3p7, abp1
          Chromosome Location:
          Chr.14: 86029335 - 86044364 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580302_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dbnl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277211.1 NP_001264140.1 1 - 2 154 64
          NM_001277212.1 NP_001264141.1 1 - 2 154 64
          NM_001277213.1 NP_001264142.1 1 - 2 154 64
          NM_031352.2 NP_112642.1 1 - 2 154 64
          GenBank mRNA AB009346.1 - 1 - 2 125 64
          AB038364.1 - 1 - 2 83 64
          AB038365.1 - 1 - 2 125 64
          AB039818.1 - 1 - 2 91 64
          AB039819.1 - 1 - 2 125 64
          BC072483.1 - 1 - 2 131 7104722 64
          FQ228854.1 - 1 - 2 154 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000935.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dbnl

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          receptor-mediated endocytosis
          synapse assembly
          Rac protein signal transduction
          neuron projection morphogenesis
          actin filament severing
          barbed-end actin filament capping
          actin filament bundle assembly
          podosome assembly
          ruffle assembly
          actin binding
          protein C-terminus binding
          protein domain specific binding
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P166494 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P149011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P149011 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P166494 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P166494 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P149011 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P166494 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P149011 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P110542 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2317 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10774001 - -
          Affymetrix Rat Gene 1.0 ST Array 10774008 - -
          Affymetrix Rat Gene 1.0 ST Array 10774010 - -
          Affymetrix Rat Gene 1.0 ST Array 10774003 - -
          Affymetrix Rat Gene 1.0 ST Array 10774014 - -
          Affymetrix Rat Gene 1.0 ST Array 10774006 - -
          Affymetrix Rat Gene 1.0 ST Array 10774012 - -
          Affymetrix Rat Gene 1.0 ST Array 10774004 - -
          Affymetrix Rat Gene 1.0 ST Array 10774009 - -
          Affymetrix Rat Gene 1.0 ST Array 10774013 - -
          Affymetrix Rat Gene 1.0 ST Array 10774005 - -
          Affymetrix Rat Gene 1.0 ST Array 10774002 - -
          Affymetrix Rat Gene 1.0 ST Array 10774007 - -
          Affymetrix Rat Gene 1.0 ST Array 10774000 - -
          Affymetrix Rat Gene 1.0 ST Array 10774011 - -
          Affymetrix Rat Gene 1.1 ST Array 10773999 - -
          Rat Genome 230 2.0 Array 1368274_at - -
          Rat Genome 230 2.0 Array 1368274_PM_at - -
          Affymetrix HT Rat Focus Array 1368274_PM_at - -
          Affymetrix HT Rat Focus Array 1368274_at - -
          Affymetrix Rat Clariom S Assay PSR1400006836.rn.1 TC1400000935.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400006839.rn.1 TC1400000935.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400006840.rn.1 TC1400000935.rn.1 RTA TC
          Rat Expression Set 230 1368274_at - -
          Rat Expression Set 230 1368274_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773999 - -
          Rat Genome U34 Set rc_AA800743_at - -
          Rat Genome U34 Set rc_AI071800_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22073990 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline