Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580421_m1
          See other IDH1 GE Assays ›
          Gene Symbol:
          Idh1
          Gene Name:
          isocitrate dehydrogenase 1 (NADP+), soluble
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 71882108 - 71911626 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580421_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Idh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031510.1 NP_113698.1 3 - 4 550 100
          XM_006245049.2 XP_006245111.1 4 - 5 608 100
          XM_008767076.1 XP_008765298.1 4 - 5 640 100
          GenBank mRNA DQ403076.1 - 3 - 4 440 100
          FQ214242.1 - 4 - 5 631 100
          L35317.1 - 3 - 4 550 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002127.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Idh1

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          glyoxylate cycle
          tricarboxylic acid cycle
          isocitrate metabolic process
          2-oxoglutarate metabolic process
          glutathione metabolic process
          response to oxidative stress
          female gonad development
          response to organic cyclic compound
          response to steroid hormone
          regulation of phospholipid catabolic process
          regulation of phospholipid biosynthetic process
          magnesium ion binding
          isocitrate dehydrogenase (NADP+) activity
          receptor binding
          protein homodimerization activity
          NADP binding
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P055450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51387 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055450 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055450 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P055450 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12569 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49789 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41495 - -
          Agilent Rat cDNA Microarray (G4105A) 701321074 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10928571 - -
          Affymetrix Rat Gene 1.0 ST Array 10928570 - -
          Affymetrix Rat Gene 1.0 ST Array 10928569 - -
          Affymetrix Rat Gene 1.0 ST Array 10928565 - -
          Affymetrix Rat Gene 1.0 ST Array 10928566 - -
          Affymetrix Rat Gene 1.0 ST Array 10928567 - -
          Affymetrix Rat Gene 1.0 ST Array 10928564 - -
          Affymetrix Rat Gene 1.0 ST Array 10928568 - -
          Affymetrix Rat Gene 1.1 ST Array 10928563 - -
          Rat Genome 230 2.0 Array 1394886_PM_at - -
          Rat Genome 230 2.0 Array 1369954_at - -
          Rat Genome 230 2.0 Array 1394886_at - -
          Rat Genome 230 2.0 Array 1369954_PM_at - -
          Affymetrix HT Rat Focus Array 1369954_PM_at - -
          Affymetrix HT Rat Focus Array 1394886_at - -
          Affymetrix HT Rat Focus Array 1369954_at - -
          Affymetrix HT Rat Focus Array 1394886_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900017487.rn.1 TC0900002127.rn.1 RTA TC
          Rat Expression Set 230 1369954_at - -
          Rat Expression Set 230 1369954_PM_at - -
          Rat Expression Set 230 1394886_PM_at - -
          Rat Expression Set 230 1394886_at - -
          Affymetrix Rat Gene 1.0 ST Array 10928563 - -
          Rat Genome U34 Set rc_AA892314_at - -
          Rat Genome U34 Set rc_AA944025_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21388288 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353960 - -
          RatRef-12 Expression BeadChip ILMN_50766 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline