Search
Search
| Gene Symbol: Nppb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031545.1 | NP_113733.1 | 2 - 3 | 402 | 106 | |
| XM_006239376.1 | XP_006239438.1 | 2 - 3 | 106 | |||
| GenBank mRNA | FQ228997.1 | - | 106 | |||
| M25297.1 | - | 2 - 3 | 402 | 106 | ||
Set Membership: |
|