Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580695_m1
          See other STAR GE Assays ›
          Gene Symbol:
          Star
          Gene Name:
          steroidogenic acute regulatory protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 71036204 - 71040847 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580695_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Star
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031558.3 NP_113746.1 3 - 4 422 79
          GenBank mRNA AB001349.1 - 378 79
          AB006007.1 - 3 - 4 423 79
          AF044081.1 - 362 79
          AY736357.1 - 3 - 4 303 79
          BC088859.1 - 3 - 4 314 7311249 79
          EF091859.1 - 2 - 3 169 79

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Star

          Gene Ontology Categories:

          Function(s) Process(es)

          organic acid metabolic process
          steroid biosynthetic process
          bile acid biosynthetic process
          progesterone biosynthetic process
          estrogen biosynthetic process
          brain development
          response to nutrient
          circadian rhythm
          cholesterol metabolic process
          glucocorticoid metabolic process
          male gonad development
          response to herbicide
          response to toxic substance
          response to organic substance
          response to ionizing radiation
          response to lead ion
          positive regulation of gene expression
          response to organic cyclic compound
          response to activity
          diterpenoid metabolic process
          response to insecticide
          insecticide metabolic process
          biphenyl metabolic process
          dibenzo-p-dioxin metabolic process
          phenol-containing compound metabolic process
          phthalate metabolic process
          response to nutrient levels
          intracellular cholesterol transport
          cellular response to insulin stimulus
          response to gonadotropin
          response to nicotine
          cellular response to interferon-alpha
          response to drug
          response to hydrogen peroxide
          circadian sleep/wake cycle, REM sleep
          response to peptide hormone
          negative regulation of neuron apoptotic process
          response to estrogen
          cellular lipid metabolic process
          response to leptin
          cellular response to fibroblast growth factor stimulus
          response to ethanol
          response to antibiotic
          regulation of neuronal synaptic plasticity
          response to steroid hormone
          positive regulation of neurogenesis
          regulation of steroid biosynthetic process
          response to corticosterone
          response to fungicide
          testosterone biosynthetic process
          cellular response to lipopolysaccharide
          cellular response to antibiotic
          cellular response to metal ion
          cellular response to cadmium ion
          cellular response to alkaloid
          cellular response to cAMP
          cellular response to glucose stimulus
          cellular response to interferon-gamma
          cellular response to gonadotropin stimulus
          cellular response to follicle-stimulating hormone stimulus
          cellular response to luteinizing hormone stimulus
          cellular response to growth hormone stimulus
          cellular response to organic cyclic compound
          cellular response to dexamethasone stimulus
          cellular response to transforming growth factor beta stimulus
          cellular response to epinephrine stimulus
          protein binding
          cholesterol binding
          cholesterol transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P154932 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P538521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408830 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538521 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154932 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154932 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538521 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P538521 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P154932 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P538521 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702558404 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700180975 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700181001 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94768 - -
          Agilent Rat cDNA Microarray (G4105A) 700181001 - -
          Agilent Rat cDNA Microarray (G4105A) 702558404 - -
          Agilent Rat cDNA Microarray (G4105A) 700180975 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792252 - -
          Affymetrix Rat Gene 1.0 ST Array 10792254 - -
          Affymetrix Rat Gene 1.0 ST Array 10792258 - -
          Affymetrix Rat Gene 1.0 ST Array 10792260 - -
          Affymetrix Rat Gene 1.0 ST Array 10792255 - -
          Affymetrix Rat Gene 1.0 ST Array 10792256 - -
          Affymetrix Rat Gene 1.0 ST Array 10792261 - -
          Affymetrix Rat Gene 1.0 ST Array 10792259 - -
          Affymetrix Rat Gene 1.0 ST Array 10792253 - -
          Affymetrix Rat Gene 1.0 ST Array 10792262 - -
          Affymetrix Rat Gene 1.0 ST Array 10792257 - -
          Affymetrix Rat Gene 1.1 ST Array 10792251 - -
          Rat Genome 230 2.0 Array 1387174_PM_a_at - -
          Rat Genome 230 2.0 Array 1368406_at - -
          Rat Genome 230 2.0 Array 1387174_a_at - -
          Rat Genome 230 2.0 Array 1389719_at - -
          Rat Genome 230 2.0 Array 1389719_PM_at - -
          Rat Genome 230 2.0 Array 1368406_PM_at - -
          Affymetrix HT Rat Focus Array 1368406_at - -
          Affymetrix HT Rat Focus Array 1368406_PM_at - -
          Affymetrix HT Rat Focus Array 1387174_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387174_a_at - -
          Rat Expression Set 230 1368406_at - -
          Rat Expression Set 230 1387174_PM_a_at - -
          Rat Expression Set 230 1368406_PM_at - -
          Rat Expression Set 230 1387174_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10792251 - -
          Rat Genome U34 Set AB001349_at - -
          Rat Genome U34 Set AB006007_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21751317 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040315 - -
          RatRef-12 Expression BeadChip ILMN_67035 - -

          Back To Top
           

          Citations (1)

          CCAAT/enhancer binding protein beta, but not steroidogenic factor-1, modulates the phthalate-induced dysregulation of rat fetal testicular steroidogenesis.
          Authors
          Kuhl AJ, Ross SM, Gaido KW
          Journal
          Endocrinology - Page No 5851 (Sat Dec 01 00:00:00 EST 2007)
          Volume (Year)
          148 (2007)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline