Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580832_m1
          See other GHRH GE Assays ›
          Gene Symbol:
          Ghrh
          Gene Name:
          growth hormone releasing hormone
          Gene Aliases:
          GHRF
          Chromosome Location:
          Chr.3: 153449124 - 153468794 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580832_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ghrh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031577.1 NP_113765.1 4 - 5 473 75
          XM_008762304.1 XP_008760526.1 4 - 5 375 75
          GenBank mRNA M73486.1 - 4 - 5 473 75
          U10156.1 - 4 - 5 375 75
          Z34092.1 - 4 - 5 375 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004609.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ghrh

          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          positive regulation of cell proliferation
          negative regulation of cardiac muscle cell apoptotic process
          cAMP-mediated signaling
          adenohypophysis development
          growth hormone secretion
          positive regulation of cAMP metabolic process
          positive regulation of cAMP biosynthetic process
          response to food
          positive regulation of multicellular organism growth
          circadian sleep/wake cycle, non-REM sleep
          response to leptin
          positive regulation of transcription, DNA-templated
          positive regulation of circadian sleep/wake cycle, REM sleep
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          positive regulation of hormone secretion
          positive regulation of growth hormone secretion
          cellular response to interleukin-6
          growth hormone-releasing hormone activity
          growth hormone-releasing hormone receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P012661 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102608 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57511 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012661 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062965 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012661 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062965 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P012661 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062965 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12596 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1048543 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701744613 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13384 - -
          Agilent Rat cDNA Microarray (G4105A) 701744613 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851332 - -
          Affymetrix Rat Gene 1.0 ST Array 10851334 - -
          Affymetrix Rat Gene 1.0 ST Array 10851328 - -
          Affymetrix Rat Gene 1.0 ST Array 10851333 - -
          Affymetrix Rat Gene 1.0 ST Array 10851331 - -
          Affymetrix Rat Gene 1.0 ST Array 10851329 - -
          Affymetrix Rat Gene 1.1 ST Array 10851327 - -
          Rat Genome 230 2.0 Array 1387217_a_at - -
          Rat Genome 230 2.0 Array 1387217_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387217_a_at - -
          Affymetrix HT Rat Focus Array 1387217_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0300035553.rn.1 TC0300004609.rn.1 RTA TC
          Rat Expression Set 230 1387217_PM_a_at - -
          Rat Expression Set 230 1387217_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851327 - -
          Rat Genome U34 Set U41183_at - -
          Rat Genome U34 Set U41183_g_at - -
          Rat Genome U34 Set Z34004exon_at - -
          Rat Genome U34 Set M73486_at - -
          Rat Genome U34 Set Z34004exon_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22368769 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352284 - -
          RatRef-12 Expression BeadChip ILMN_60128 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline