Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580893_m1
          See other SLC22A2 GE Assays ›
          Gene Symbol:
          Slc22a2
          Gene Name:
          solute carrier family 22 (organic cation transporter), member 2
          Gene Aliases:
          OCT2, OCT2r, rOCT2
          Chromosome Location:
          Chr.1: 48318025 - 48360219 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580893_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc22a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031584.2 NP_113772.1 9 - 10 1592 55
          GenBank mRNA D83044.1 - 9 - 10 1595 55
          X98334.1 - 9 - 10 1541 55
          Y13154.1 - 9 - 10 1542 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004457.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc22a2

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          organic cation transport
          quaternary ammonium group transport
          choline transport
          ion transmembrane transport
          histamine transport
          cation transmembrane transport
          steroid binding
          ion transmembrane transporter activity
          organic cation transmembrane transporter activity
          choline transmembrane transporter activity
          quaternary ammonium group transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12866 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145979 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166278 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078679 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078679 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078679 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P266976 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701830325 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701829374 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18106 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13173 - -
          Agilent Rat cDNA Microarray (G4105A) 701829374 - -
          Agilent Rat cDNA Microarray (G4105A) 701830325 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10717959 - -
          Affymetrix Rat Gene 1.0 ST Array 10717957 - -
          Affymetrix Rat Gene 1.0 ST Array 10717964 - -
          Affymetrix Rat Gene 1.0 ST Array 10717961 - -
          Affymetrix Rat Gene 1.0 ST Array 10717960 - -
          Affymetrix Rat Gene 1.0 ST Array 10717956 - -
          Affymetrix Rat Gene 1.0 ST Array 10717966 - -
          Affymetrix Rat Gene 1.0 ST Array 10717963 - -
          Affymetrix Rat Gene 1.0 ST Array 10717965 - -
          Affymetrix Rat Gene 1.0 ST Array 10717962 - -
          Affymetrix Rat Gene 1.0 ST Array 10717958 - -
          Affymetrix Rat Gene 1.1 ST Array 10717955 - -
          Rat Genome 230 2.0 Array 1387303_PM_at - -
          Rat Genome 230 2.0 Array 1387303_at - -
          Affymetrix HT Rat Focus Array 1387303_PM_at - -
          Affymetrix HT Rat Focus Array 1387303_at - -
          Affymetrix Rat Clariom S Assay PSR0100035405.rn.1 TC0100004457.rn.1 RTA TC
          Rat Expression Set 230 1387303_at - -
          Rat Expression Set 230 1387303_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10717955 - -
          Rat Genome U34 Set D83044_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21329405 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361093 - -
          RatRef-12 Expression BeadChip ILMN_60219 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline