Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580979_m1
          See other SART1 GE Assays ›
          Gene Symbol:
          Sart1
          Gene Name:
          squamous cell carcinoma antigen recognized by T cells
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 220762496 - 220771158 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580979_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sart1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031596.1 NP_113784.1 3 - 4 487 56
          GenBank mRNA AB014722.1 - 3 - 4 487 56
          BC083551.1 - 3 - 4 460 7134496 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007111.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sart1

          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal snRNP assembly
          maturation of 5S rRNA
          mRNA cis splicing, via spliceosome
          positive regulation of cytotoxic T cell differentiation
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P177742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167592 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P177742 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P177742 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P177742 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P177742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55789 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727822 - -
          Affymetrix Rat Gene 1.0 ST Array 10727820 - -
          Affymetrix Rat Gene 1.0 ST Array 10727816 - -
          Affymetrix Rat Gene 1.0 ST Array 10727819 - -
          Affymetrix Rat Gene 1.0 ST Array 10727834 - -
          Affymetrix Rat Gene 1.0 ST Array 10727827 - -
          Affymetrix Rat Gene 1.0 ST Array 10727824 - -
          Affymetrix Rat Gene 1.0 ST Array 10727823 - -
          Affymetrix Rat Gene 1.0 ST Array 10727817 - -
          Affymetrix Rat Gene 1.0 ST Array 10727831 - -
          Affymetrix Rat Gene 1.0 ST Array 10727829 - -
          Affymetrix Rat Gene 1.0 ST Array 10713041 - -
          Affymetrix Rat Gene 1.0 ST Array 10727821 - -
          Affymetrix Rat Gene 1.0 ST Array 10727832 - -
          Affymetrix Rat Gene 1.0 ST Array 10727828 - -
          Affymetrix Rat Gene 1.0 ST Array 10727830 - -
          Affymetrix Rat Gene 1.0 ST Array 10727826 - -
          Affymetrix Rat Gene 1.0 ST Array 10727833 - -
          Affymetrix Rat Gene 1.0 ST Array 10727825 - -
          Affymetrix Rat Gene 1.0 ST Array 10727835 - -
          Affymetrix Rat Gene 1.0 ST Array 10727818 - -
          Affymetrix Rat Gene 1.1 ST Array 10713040 - -
          Affymetrix Rat Gene 1.1 ST Array 10727815 - -
          Rat Genome 230 2.0 Array 1387012_at - -
          Rat Genome 230 2.0 Array 1387012_PM_at - -
          Affymetrix HT Rat Focus Array 1387012_at - -
          Affymetrix HT Rat Focus Array 1387012_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100056824.rn.1 TC0100007111.rn.1 RTA TC
          Rat Expression Set 230 1387012_PM_at - -
          Rat Expression Set 230 1387012_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727815 - -
          Rat Genome U34 Set AB014722_at - -
          Rat Genome U34 Set AB014722_g_at - -
          Rat Genome U34 Set rc_AI228721_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21912102 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353310 - -
          RatRef-12 Expression BeadChip ILMN_51397 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline