Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581074_m1
          See other ATP6V0A1 GE Assays ›
          Gene Symbol:
          Atp6v0a1
          Gene Name:
          ATPase, H+ transporting, lysosomal V0 subunit A1
          Gene Aliases:
          Atp6n1, Atp6n1a
          Chromosome Location:
          Chr.10: 88914275 - 88967736 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581074_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp6v0a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031604.2 NP_113792.2 10 - 11 1182 105
          XM_006247295.2 XP_006247357.1 10 - 11 1203 105
          XM_006247296.2 XP_006247358.1 10 - 11 1203 105
          XM_006247297.2 XP_006247359.1 10 - 11 1182 105
          XM_008768002.1 XP_008766224.1 10 - 11 1166 105
          XM_008768003.1 XP_008766225.1 10 - 11 1145 105
          XM_008768004.1 XP_008766226.1 10 - 11 1204 105
          GenBank mRNA DQ286421.1 - 9 - 10 1024 105
          DQ286422.1 - 9 - 10 1045 105
          DQ286423.1 - 9 - 10 1045 105
          DQ286424.1 - 9 - 10 1024 105
          M58758.1 - 9 - 10 1054 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001642.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp6v0a1

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          vacuolar acidification
          biological_process
          ATP synthesis coupled proton transport
          ATP hydrolysis coupled proton transport
          vacuolar proton-transporting V-type ATPase complex assembly
          toxin transport
          molecular_function
          hydrogen ion transmembrane transporter activity
          proton-transporting ATPase activity, rotational mechanism
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P002989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884565 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118585 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146236 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74749 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91930 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002989 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P002989 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P402760 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P293791 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701314903 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68947 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94712 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79976 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38513 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3824 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59134 - -
          Agilent Rat cDNA Microarray (G4105A) 701314903 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10738248 - -
          Affymetrix Rat Gene 1.0 ST Array 10738250 - -
          Affymetrix Rat Gene 1.0 ST Array 10738256 - -
          Affymetrix Rat Gene 1.0 ST Array 10738241 - -
          Affymetrix Rat Gene 1.0 ST Array 10738254 - -
          Affymetrix Rat Gene 1.0 ST Array 10738253 - -
          Affymetrix Rat Gene 1.0 ST Array 10738246 - -
          Affymetrix Rat Gene 1.0 ST Array 10738257 - -
          Affymetrix Rat Gene 1.0 ST Array 10738239 - -
          Affymetrix Rat Gene 1.0 ST Array 10738255 - -
          Affymetrix Rat Gene 1.0 ST Array 10738245 - -
          Affymetrix Rat Gene 1.0 ST Array 10738258 - -
          Affymetrix Rat Gene 1.0 ST Array 10738252 - -
          Affymetrix Rat Gene 1.0 ST Array 10738238 - -
          Affymetrix Rat Gene 1.0 ST Array 10738242 - -
          Affymetrix Rat Gene 1.0 ST Array 10738243 - -
          Affymetrix Rat Gene 1.0 ST Array 10738244 - -
          Affymetrix Rat Gene 1.0 ST Array 10738251 - -
          Affymetrix Rat Gene 1.0 ST Array 10738240 - -
          Affymetrix Rat Gene 1.0 ST Array 10738259 - -
          Affymetrix Rat Gene 1.1 ST Array 10738237 - -
          Rat Genome 230 2.0 Array 1387045_PM_at - -
          Rat Genome 230 2.0 Array 1387045_at - -
          Affymetrix HT Rat Focus Array 1387045_PM_at - -
          Affymetrix HT Rat Focus Array 1387045_at - -
          Affymetrix Rat Clariom S Assay PSR1000014559.rn.1 TC1000001642.rn.1 RTA TC
          Rat Expression Set 230 1387045_at - -
          Rat Expression Set 230 1387045_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10738237 - -
          Rat Genome U34 Set M58758_g_at - -
          Rat Genome U34 Set M58758_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20801059 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358792 - -
          RatRef-12 Expression BeadChip ILMN_59883 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline