Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581100_m1
          See other TMOD2 GE Assays ›
          Gene Symbol:
          Tmod2
          Gene Name:
          tropomodulin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 82505947 - 82547569 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581100_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmod2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031613.1 NP_113801.1 6 - 7 790 103
          XM_006243407.2 XP_006243469.1 7 - 8 1250 103
          XM_006243408.2 XP_006243470.1 7 - 8 4452 103
          XM_008766331.1 XP_008764553.1 7 - 8 842 103
          GenBank mRNA U59240.1 - 6 - 7 790 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002945.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmod2

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          actin filament organization
          neuron-neuron synaptic transmission
          learning or memory
          myofibril assembly
          positive regulation of G-protein coupled receptor protein signaling pathway
          pointed-end actin filament capping
          actin binding
          tropomyosin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40259 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54582 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12614 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12614 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12614 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82109 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27704 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918732 - -
          Affymetrix Rat Gene 1.0 ST Array 10918731 - -
          Affymetrix Rat Gene 1.0 ST Array 10918734 - -
          Affymetrix Rat Gene 1.0 ST Array 10918730 - -
          Affymetrix Rat Gene 1.0 ST Array 10918729 - -
          Affymetrix Rat Gene 1.0 ST Array 10918737 - -
          Affymetrix Rat Gene 1.0 ST Array 10918735 - -
          Affymetrix Rat Gene 1.0 ST Array 10918736 - -
          Affymetrix Rat Gene 1.0 ST Array 10918733 - -
          Affymetrix Rat Gene 1.1 ST Array 10918728 - -
          Rat Genome 230 2.0 Array 1390482_PM_at - -
          Rat Genome 230 2.0 Array 1393418_PM_at - -
          Rat Genome 230 2.0 Array 1369541_at - -
          Rat Genome 230 2.0 Array 1369541_PM_at - -
          Rat Genome 230 2.0 Array 1390482_at - -
          Rat Genome 230 2.0 Array 1393418_at - -
          Affymetrix HT Rat Focus Array 1393418_PM_at - -
          Affymetrix HT Rat Focus Array 1369541_at - -
          Affymetrix HT Rat Focus Array 1369541_PM_at - -
          Affymetrix HT Rat Focus Array 1393418_at - -
          Affymetrix Rat Clariom S Assay PSR0800023639.rn.1 TC0800002945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023638.rn.1 TC0800002945.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023640.rn.1 TC0800002945.rn.1 RTA TC
          Rat Expression Set 230 1393418_PM_at - -
          Rat Expression Set 230 1369541_PM_at - -
          Rat Expression Set 230 1369541_at - -
          Rat Expression Set 230 1393418_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918728 - -
          Rat Genome U34 Set U59240_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20874227 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362257 - -
          RatRef-12 Expression BeadChip ILMN_49399 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline