Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581128_m1
          See other ZRANB2 GE Assays ›
          Gene Symbol:
          Zranb2
          Gene Name:
          zinc finger, RAN-binding domain containing 2
          Gene Aliases:
          Zfp265, Zis, Znf265
          Chromosome Location:
          Chr.2: 263864331 - 263877821 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581128_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zranb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031616.1 NP_113804.1 9 - 10 946 100
          GenBank mRNA AF013965.1 - 9 - 10 1186 100
          AF013967.1 - 9 - 10 946 100
          BC087012.1 - 9 - 10 933 7189430 101
          FQ211774.1 - 9 - 10 973 101
          FQ211917.1 - 9 - 10 925 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002455.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zranb2

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          zinc ion binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172503 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068801 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068801 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068801 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P359618 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P608365 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702023331 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30319 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19247 - -
          Agilent Rat cDNA Microarray (G4105A) 702023331 - -
          Agilent Rat cDNA Microarray (G4105A) 700280327 - -
          Agilent Rat cDNA Microarray (G4105A) 700292320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819898 - -
          Affymetrix Rat Gene 1.0 ST Array 10819893 - -
          Affymetrix Rat Gene 1.0 ST Array 10819901 - -
          Affymetrix Rat Gene 1.0 ST Array 10819899 - -
          Affymetrix Rat Gene 1.0 ST Array 10819896 - -
          Affymetrix Rat Gene 1.0 ST Array 10819900 - -
          Affymetrix Rat Gene 1.0 ST Array 10819892 - -
          Affymetrix Rat Gene 1.0 ST Array 10819891 - -
          Affymetrix Rat Gene 1.0 ST Array 10819902 - -
          Affymetrix Rat Gene 1.0 ST Array 10819895 - -
          Affymetrix Rat Gene 1.0 ST Array 10819897 - -
          Affymetrix Rat Gene 1.0 ST Array 10819894 - -
          Affymetrix Rat Gene 1.1 ST Array 10819890 - -
          Rat Genome 230 2.0 Array 1368228_PM_at - -
          Rat Genome 230 2.0 Array 1387117_at - -
          Rat Genome 230 2.0 Array 1387117_PM_at - -
          Rat Genome 230 2.0 Array 1368228_at - -
          Affymetrix HT Rat Focus Array 1387117_PM_at - -
          Affymetrix HT Rat Focus Array 1368228_PM_at - -
          Affymetrix HT Rat Focus Array 1368228_at - -
          Affymetrix HT Rat Focus Array 1387117_at - -
          Affymetrix Rat Clariom S Assay PSR0200017213.rn.1 TC0200002455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017216.rn.1 TC0200002455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017212.rn.1 TC0200002455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017214.rn.1 TC0200002455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017211.rn.1 TC0200002455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017215.rn.1 TC0200002455.rn.1 RTA TC
          Rat Expression Set 230 1368228_PM_at - -
          Rat Expression Set 230 1387117_PM_at - -
          Rat Expression Set 230 1368228_at - -
          Rat Expression Set 230 1387117_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819890 - -
          Rat Genome U34 Set rc_AI170608_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21836817 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359067 - -
          RatRef-12 Expression BeadChip ILMN_56901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline