Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581130_m1
          See other CSNK1E GE Assays ›
          Gene Symbol:
          Csnk1e
          Gene Name:
          casein kinase 1, epsilon
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 120651976 - 120675559 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581130_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csnk1e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031617.1 NP_113805.1 3 - 4 337 90
          XM_006242038.2 XP_006242100.1 4 - 5 486 90
          GenBank mRNA AB042191.1 - 3 - 4 337 90
          AB042192.1 - 3 - 4 337 90
          AB056113.1 - 3 - 4 337 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003497.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csnk1e

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          endocytosis
          regulation of cell shape
          Wnt signaling pathway
          peptidyl-serine phosphorylation
          negative regulation of Wnt signaling pathway
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          circadian regulation of gene expression
          cellular protein localization
          regulation of circadian rhythm
          positive regulation of canonical Wnt signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P127944 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P515454 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P040498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170708 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10662 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125583 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P515454 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127944 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P515454 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127944 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P515454 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P127944 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P040498 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P192988 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P515454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42940 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48999 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10905443 - -
          Affymetrix Rat Gene 1.0 ST Array 10905452 - -
          Affymetrix Rat Gene 1.0 ST Array 10905441 - -
          Affymetrix Rat Gene 1.0 ST Array 10905451 - -
          Affymetrix Rat Gene 1.0 ST Array 10905442 - -
          Affymetrix Rat Gene 1.0 ST Array 10905448 - -
          Affymetrix Rat Gene 1.0 ST Array 10905450 - -
          Affymetrix Rat Gene 1.0 ST Array 10905447 - -
          Affymetrix Rat Gene 1.0 ST Array 10905449 - -
          Affymetrix Rat Gene 1.1 ST Array 10905439 - -
          Rat Genome 230 2.0 Array 1386693_PM_at - -
          Rat Genome 230 2.0 Array 1384161_PM_at - -
          Rat Genome 230 2.0 Array 1384161_at - -
          Rat Genome 230 2.0 Array 1386693_at - -
          Rat Genome 230 2.0 Array 1369804_PM_a_at - -
          Rat Genome 230 2.0 Array 1369804_a_at - -
          Affymetrix HT Rat Focus Array 1386693_PM_at - -
          Affymetrix HT Rat Focus Array 1369804_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369804_a_at - -
          Affymetrix HT Rat Focus Array 1384161_at - -
          Affymetrix HT Rat Focus Array 1384161_PM_at - -
          Affymetrix HT Rat Focus Array 1386693_at - -
          Affymetrix Rat Clariom S Assay PSR0700027512.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027508.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027510.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027521.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027498.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027501.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027500.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027499.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027509.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027513.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027511.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027514.rn.1 TC0700003497.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027520.rn.1 TC0700003497.rn.1 RTA TC
          Rat Expression Set 230 1369804_PM_a_at - -
          Rat Expression Set 230 1369804_a_at - -
          Rat Expression Set 230 1386693_PM_at - -
          Rat Expression Set 230 1386693_at - -
          Rat Expression Set 230 1384161_PM_at - -
          Rat Expression Set 230 1384161_at - -
          Affymetrix Rat Gene 1.0 ST Array 10905439 - -
          Rat Genome U34 Set rc_AA996676_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21952575 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline