Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581196_g1
          See other PSMB4 GE Assays ›
          Gene Symbol:
          Psmb4
          Gene Name:
          proteasome (prosome, macropain) subunit, beta type 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 196043618 - 196046311 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581196_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmb4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031629.1 NP_113817.1 6 - 7 784 67
          GenBank mRNA FQ215475.1 - 6 - 7 802 67
          FQ216787.1 - 6 - 7 801 67
          FQ218364.1 - 6 - 7 803 67
          FQ219781.1 - 6 - 7 802 67
          FQ220578.1 - 6 - 7 801 67
          FQ220909.1 - 6 - 7 803 67
          FQ225189.1 - 6 - 7 841 67
          FQ226073.1 - 6 - 7 802 67
          FQ226711.1 - 6 - 7 802 67
          FQ228014.1 - 6 - 7 802 67
          FQ232850.1 - 6 - 7 803 67
          L17127.1 - 6 - 7 784 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004187.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmb4

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of inflammatory response to antigenic stimulus
          proteolysis involved in cellular protein catabolic process
          lipopolysaccharide binding
          threonine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P099189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924116 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099189 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099189 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P099189 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P477905 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700525466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18645 - -
          Agilent Rat cDNA Microarray (G4105A) 700525466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824887 - -
          Affymetrix Rat Gene 1.0 ST Array 10824888 - -
          Affymetrix Rat Gene 1.0 ST Array 10824886 - -
          Affymetrix Rat Gene 1.0 ST Array 10824889 - -
          Affymetrix Rat Gene 1.0 ST Array 10824890 - -
          Affymetrix Rat Gene 1.0 ST Array 10824891 - -
          Affymetrix Rat Gene 1.1 ST Array 10824884 - -
          Rat Genome 230 2.0 Array 1398831_at - -
          Rat Genome 230 2.0 Array 1398831_PM_at - -
          Affymetrix HT Rat Focus Array 1398831_at - -
          Affymetrix HT Rat Focus Array 1398831_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200028560.rn.1 TC0200004187.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028561.rn.1 TC0200004187.rn.1 RTA TC
          Rat Expression Set 230 1398831_at - -
          Rat Expression Set 230 1398831_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824884 - -
          Rat Genome U34 Set L17127_at - -
          Rat Genome U34 Set L17127_g_at - -
          Rat Genome U34 Set rc_AI172162_at - -
          Rat Genome U34 Set L17127_at - -
          Rat Genome U34 Set L17127_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21469292 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371481 - -
          RatRef-12 Expression BeadChip ILMN_64689 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline