Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581297_m1
          See other SFMBT1 GE Assays ›
          Gene Symbol:
          Sfmbt1
          Gene Name:
          Scm-like with four mbt domains 1
          Gene Aliases:
          Sfmbt
          Chromosome Location:
          Chr.16: 6775648 - 6899669 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581297_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sfmbt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031647.2 NP_113835.2 9 - 10 1248 80
          XM_006252656.2 XP_006252718.1 9 - 10 1534 80
          XM_006252657.2 XP_006252719.1 10 - 11 1594 80
          XM_006252658.1 XP_006252720.1 9 - 10 3506 80
          XM_008771003.1 XP_008769225.1 11 - 12 2515 80
          XM_008771004.1 XP_008769226.1 10 - 11 2455 80
          XM_008771005.1 XP_008769227.1 12 - 13 1824 80
          XM_008771006.1 XP_008769228.1 11 - 12 1688 80
          GenBank mRNA AB032164.1 - 9 - 10 1242 80
          BC081770.1 - 9 - 10 1248 7132495 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000083.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sfmbt1

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          spermatogenesis
          chromatin modification
          cell differentiation
          negative regulation of transcription, DNA-templated
          negative regulation of muscle organ development
          histone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P465408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549883 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128606 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71679 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179077 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P465408 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P465408 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P465408 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P465408 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701287267 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700503807 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701942630 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51407 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55198 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53293 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83405 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5144 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18074 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26791 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47518 - -
          Agilent Rat cDNA Microarray (G4105A) 700503807 - -
          Agilent Rat cDNA Microarray (G4105A) 701287267 - -
          Agilent Rat cDNA Microarray (G4105A) 701942630 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786511 - -
          Affymetrix Rat Gene 1.0 ST Array 10786496 - -
          Affymetrix Rat Gene 1.0 ST Array 10786506 - -
          Affymetrix Rat Gene 1.0 ST Array 10786501 - -
          Affymetrix Rat Gene 1.0 ST Array 10786503 - -
          Affymetrix Rat Gene 1.0 ST Array 10786512 - -
          Affymetrix Rat Gene 1.0 ST Array 10786514 - -
          Affymetrix Rat Gene 1.0 ST Array 10786502 - -
          Affymetrix Rat Gene 1.0 ST Array 10786505 - -
          Affymetrix Rat Gene 1.0 ST Array 10786500 - -
          Affymetrix Rat Gene 1.0 ST Array 10786516 - -
          Affymetrix Rat Gene 1.0 ST Array 10786507 - -
          Affymetrix Rat Gene 1.0 ST Array 10786515 - -
          Affymetrix Rat Gene 1.0 ST Array 10786508 - -
          Affymetrix Rat Gene 1.0 ST Array 10786499 - -
          Affymetrix Rat Gene 1.0 ST Array 10786510 - -
          Affymetrix Rat Gene 1.0 ST Array 10786497 - -
          Affymetrix Rat Gene 1.0 ST Array 10786509 - -
          Affymetrix Rat Gene 1.0 ST Array 10786504 - -
          Affymetrix Rat Gene 1.0 ST Array 10786498 - -
          Affymetrix Rat Gene 1.0 ST Array 10786513 - -
          Affymetrix Rat Gene 1.1 ST Array 10786495 - -
          Rat Genome 230 2.0 Array 1368129_at - -
          Rat Genome 230 2.0 Array 1368129_PM_at - -
          Affymetrix HT Rat Focus Array 1368129_PM_at - -
          Affymetrix HT Rat Focus Array 1368129_at - -
          Affymetrix Rat Clariom S Assay PSR1600000760.rn.1 TC1600000083.rn.1 RTA TC
          Rat Expression Set 230 1368129_at - -
          Rat Expression Set 230 1368129_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786495 - -
          Rat Genome U34 Set rc_AA893965_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21396215 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353884 - -
          RatRef-12 Expression BeadChip ILMN_58881 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline