Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581299_m1
          See other FXYD1 GE Assays ›
          Gene Symbol:
          Fxyd1
          Gene Name:
          FXYD domain-containing ion transport regulator 1
          Gene Aliases:
          Plm
          Chromosome Location:
          Chr.1: 89484197 - 89488278 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581299_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fxyd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031648.2 NP_113836.2 5 - 6 286 70
          XM_006228838.2 XP_006228900.1 6 - 7 506 70
          XM_006228839.2 XP_006228901.1 5 - 6 281 70
          XM_008759184.1 XP_008757406.1 5 - 6 295 70
          GenBank mRNA CB712570.1 - 5 - 6 286 70
          FQ211349.1 - 5 - 6 297 70
          FQ224131.1 - 5 - 6 249 70
          FQ224338.1 - 5 - 6 332 70
          U72246.1 - 4 - 5 200 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005182.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fxyd1

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          brain development
          positive regulation of organic acid transport
          regulation of cytosolic calcium ion concentration
          regulation of cardiac muscle cell contraction
          regulation of cardiac muscle cell membrane potential
          chloride transmembrane transport
          positive regulation of sodium ion export from cell
          positive regulation of inorganic anion transmembrane transport
          regulation of sodium ion transmembrane transporter activity
          chloride channel activity
          protein binding
          myosin binding
          sodium channel regulator activity
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P572123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287983 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P572123 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P572123 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P572123 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P572123 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720882 - -
          Affymetrix Rat Gene 1.0 ST Array 10720880 - -
          Affymetrix Rat Gene 1.0 ST Array 10720881 - -
          Affymetrix Rat Gene 1.0 ST Array 10720883 - -
          Affymetrix Rat Gene 1.0 ST Array 10720879 - -
          Affymetrix Rat Gene 1.1 ST Array 10720878 - -
          Rat Genome 230 2.0 Array 1369960_PM_at - -
          Rat Genome 230 2.0 Array 1369960_at - -
          Affymetrix HT Rat Focus Array 1369960_PM_at - -
          Affymetrix HT Rat Focus Array 1369960_at - -
          Affymetrix Rat Clariom S Assay PSR0100041828.rn.1 TC0100005182.rn.1 RTA TC
          Rat Expression Set 230 1369960_PM_at - -
          Rat Expression Set 230 1369960_at - -
          Affymetrix Rat Gene 1.0 ST Array 10720878 - -
          Rat Genome U34 Set rc_AA799645_at - -
          Rat Genome U34 Set rc_AA799645_g_at - -
          Rat Genome U34 Set rc_AI009344_i_at - -
          Rat Genome U34 Set rc_AA926363_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21707297 - -
          ABI Rat Genome Survey Microarray 22115468 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372940 - -
          RatRef-12 Expression BeadChip ILMN_62914 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline