Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581408_m1
          See other TGM1 GE Assays ›
          Gene Symbol:
          Tgm1
          Gene Name:
          transglutaminase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 34378136 - 34393156 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581408_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031659.1 NP_113847.1 9 - 10 1531 87
          XM_006252014.2 XP_006252076.1 9 - 10 3078 87
          XM_008770691.1 XP_008768913.1 9 - 10 3169 87
          GenBank mRNA BC097305.1 - 9 - 10 1487 7462186 87
          FQ210559.1 - 9 - 10 1536 87
          M57263.1 - 9 - 10 1531 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001719.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgm1

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          organ morphogenesis
          peptide cross-linking
          keratinocyte differentiation
          keratinization
          protein-glutamine gamma-glutamyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P399249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P399249 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P399249 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P399249 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P399249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98504 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16495 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76038 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783883 - -
          Affymetrix Rat Gene 1.0 ST Array 10783886 - -
          Affymetrix Rat Gene 1.0 ST Array 10783893 - -
          Affymetrix Rat Gene 1.0 ST Array 10783892 - -
          Affymetrix Rat Gene 1.0 ST Array 10783885 - -
          Affymetrix Rat Gene 1.0 ST Array 10783882 - -
          Affymetrix Rat Gene 1.0 ST Array 10783881 - -
          Affymetrix Rat Gene 1.0 ST Array 10783888 - -
          Affymetrix Rat Gene 1.0 ST Array 10783891 - -
          Affymetrix Rat Gene 1.0 ST Array 10783890 - -
          Affymetrix Rat Gene 1.0 ST Array 10783887 - -
          Affymetrix Rat Gene 1.0 ST Array 10783884 - -
          Affymetrix Rat Gene 1.0 ST Array 10783896 - -
          Affymetrix Rat Gene 1.0 ST Array 10783889 - -
          Affymetrix Rat Gene 1.0 ST Array 10783895 - -
          Affymetrix Rat Gene 1.0 ST Array 10783894 - -
          Affymetrix Rat Gene 1.1 ST Array 10783880 - -
          Rat Genome 230 2.0 Array 1370051_at - -
          Rat Genome 230 2.0 Array 1370051_PM_at - -
          Affymetrix HT Rat Focus Array 1370051_PM_at - -
          Affymetrix HT Rat Focus Array 1370051_at - -
          Affymetrix Rat Clariom S Assay PSR1500011183.rn.1 TC1500001719.rn.1 RTA TC
          Rat Expression Set 230 1370051_PM_at - -
          Rat Expression Set 230 1370051_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783880 - -
          Rat Genome U34 Set M57263_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21654269 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360947 - -
          RatRef-12 Expression BeadChip ILMN_61579 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0