Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581785_m1
          See other HOMER1 GE Assays ›
          Gene Symbol:
          Homer1
          Gene Name:
          homer homolog 1 (Drosophila)
          Gene Aliases:
          HOMER1F, Vesl-1
          Chromosome Location:
          Chr.2: 22909554 - 23012303 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581785_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Homer1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031707.1 NP_113895.1 7 - 8 1011 83
          XM_006231775.2 XP_006231837.1 6 - 7 1556 83
          XM_008760635.1 XP_008758857.1 7 - 8 1579 83
          GenBank mRNA AB007688.1 - 7 - 8 1018 83
          AB017140.1 - 7 - 8 1011 83
          AF093267.1 - 6 - 7 1420 83
          AF093268.1 - 7 - 8 1448 83
          AJ276328.1 - 7 - 8 731 83
          AY189942.1 - 2 - 3 354 83
          AY189943.1 - 3 - 4 390 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000158.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Homer1

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          G-protein coupled glutamate receptor signaling pathway
          circadian rhythm
          response to nicotine
          protein localization to synapse
          response to cocaine
          behavioral response to cocaine
          skeletal muscle fiber development
          chemical homeostasis within a tissue
          response to calcium ion
          positive regulation of calcium ion transport
          regulation of calcium ion import
          regulation of store-operated calcium entry
          regulation of cation channel activity
          receptor binding
          protein binding
          type 5 metabotropic glutamate receptor binding
          protein complex scaffold
          G-protein coupled glutamate receptor binding
          identical protein binding
          ion channel binding
          protein heterodimerization activity
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P764228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38765 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P764228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P764228 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P764228 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P148865 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P380835 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702394036 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701030467 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71177 - -
          Agilent Rat cDNA Microarray (G4105A) 701030467 - -
          Agilent Rat cDNA Microarray (G4105A) 702394036 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940541 - -
          Affymetrix Rat Gene 1.1 ST Array 10940541 - -
          Rat Genome 230 2.0 Array 1368319_PM_a_at - -
          Rat Genome 230 2.0 Array 1399163_a_at - -
          Rat Genome 230 2.0 Array 1370454_PM_at - -
          Rat Genome 230 2.0 Array 1370997_at - -
          Rat Genome 230 2.0 Array 1370997_PM_at - -
          Rat Genome 230 2.0 Array 1370656_PM_a_at - -
          Rat Genome 230 2.0 Array 1399163_PM_a_at - -
          Rat Genome 230 2.0 Array 1370656_a_at - -
          Rat Genome 230 2.0 Array 1368318_PM_at - -
          Rat Genome 230 2.0 Array 1368318_at - -
          Rat Genome 230 2.0 Array 1370454_at - -
          Rat Genome 230 2.0 Array 1368319_a_at - -
          Affymetrix HT Rat Focus Array 1368319_a_at - -
          Affymetrix HT Rat Focus Array 1368318_PM_at - -
          Affymetrix HT Rat Focus Array 1399163_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368319_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370997_at - -
          Affymetrix HT Rat Focus Array 1370656_a_at - -
          Affymetrix HT Rat Focus Array 1370454_at - -
          Affymetrix HT Rat Focus Array 1399163_a_at - -
          Affymetrix HT Rat Focus Array 1368318_at - -
          Affymetrix HT Rat Focus Array 1370454_PM_at - -
          Affymetrix HT Rat Focus Array 1370997_PM_at - -
          Affymetrix HT Rat Focus Array 1370656_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0200000818.rn.1 TC0200000158.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000816.rn.1 TC0200000158.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000817.rn.1 TC0200000158.rn.1 RTA TC
          Rat Expression Set 230 1370454_PM_at - -
          Rat Expression Set 230 1368319_PM_a_at - -
          Rat Expression Set 230 1368318_at - -
          Rat Expression Set 230 1370656_a_at - -
          Rat Expression Set 230 1370997_PM_at - -
          Rat Expression Set 230 1370656_PM_a_at - -
          Rat Expression Set 230 1399163_a_at - -
          Rat Expression Set 230 1370997_at - -
          Rat Expression Set 230 1368318_PM_at - -
          Rat Expression Set 230 1368319_a_at - -
          Rat Expression Set 230 1399163_PM_a_at - -
          Rat Expression Set 230 1370454_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940541 - -
          Rat Genome U34 Set AF093267_s_at - -
          Rat Genome U34 Set AB017140_at - -
          Rat Genome U34 Set rc_AA799735_g_at - -
          Rat Genome U34 Set AF093268_s_at - -
          Rat Genome U34 Set AB007688_s_at - -
          Rat Genome U34 Set AB017140_g_at - -
          Rat Genome U34 Set AB003726_at - -
          Rat Genome U34 Set AF030088UTR#1_at - -
          Rat Genome U34 Set rc_AA925598_at - -
          Rat Genome U34 Set AF030088UTR#1_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21726000 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline