Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581820_m1
          See other PDZK1 GE Assays ›
          Gene Symbol:
          Pdzk1
          Gene Name:
          PDZ domain containing 1
          Gene Aliases:
          Clamp
          Chromosome Location:
          Chr.2: 198965685 - 198999323 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581820_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdzk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031712.1 NP_113900.1 6 - 7 1061 61
          GenBank mRNA AF013145.1 - 6 - 7 1035 61
          AF116896.1 - 6 - 7 1061 61
          BC078788.1 - 6 - 7 1053 7109724 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001776.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdzk1

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          carnitine transport
          cholesterol transport
          positive regulation of ion transmembrane transport
          regulation of anion transport
          intracellular transport
          establishment of protein localization to plasma membrane
          positive regulation of protein targeting to membrane
          scavenger receptor binding
          protein binding
          PDZ domain binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P222186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2830 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160791 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40449 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P222186 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P222186 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P222186 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P222186 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65798 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90187 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97614 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10817659 - -
          Affymetrix Rat Gene 1.0 ST Array 10817661 - -
          Affymetrix Rat Gene 1.0 ST Array 10817662 - -
          Affymetrix Rat Gene 1.0 ST Array 10817660 - -
          Affymetrix Rat Gene 1.0 ST Array 10817658 - -
          Affymetrix Rat Gene 1.0 ST Array 10817663 - -
          Affymetrix Rat Gene 1.0 ST Array 10817664 - -
          Affymetrix Rat Gene 1.0 ST Array 10817665 - -
          Affymetrix Rat Gene 1.0 ST Array 10817657 - -
          Affymetrix Rat Gene 1.1 ST Array 10817656 - -
          Rat Genome 230 2.0 Array 1368178_PM_at - -
          Rat Genome 230 2.0 Array 1368178_at - -
          Affymetrix HT Rat Focus Array 1368178_PM_at - -
          Affymetrix HT Rat Focus Array 1368178_at - -
          Affymetrix Rat Clariom S Assay PSR0200012125.rn.1 TC0200001776.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200012124.rn.1 TC0200001776.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200012122.rn.1 TC0200001776.rn.1 RTA TC
          Rat Expression Set 230 1368178_at - -
          Rat Expression Set 230 1368178_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10817656 - -
          Rat Genome U34 Set AF013145_at - -
          Rat Genome U34 Set rc_AI232999_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21270725 - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_61185 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline