Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581848_m1
          See other PFKM GE Assays ›
          Gene Symbol:
          Pfkm
          Gene Name:
          phosphofructokinase, muscle
          Gene Aliases:
          ATP-PFK, PFK-A, Pfk-M
          Chromosome Location:
          Chr.7: 139702066 - 139722132 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581848_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pfkm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031715.1 NP_113903.1 275 57
          GenBank mRNA BC094212.1 - 3 - 4 262 7314667 57
          D21869.1 - 3 - 4 219 57
          FN433010.1 - 3 - 4 240 57
          U25651.1 - 275 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001766.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pfkm

          Panther Classification:

          Molecular Function -

          carbohydrate kinase kinase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular glucose homeostasis
          fructose 6-phosphate metabolic process
          glycolytic process
          fructose 1,6-bisphosphate metabolic process
          positive regulation of insulin secretion
          glucose homeostasis
          muscle cell cellular homeostasis
          carbohydrate phosphorylation
          protein oligomerization
          protein homotetramerization
          6-phosphofructokinase activity
          ATP binding
          protein C-terminus binding
          phosphofructokinase activity
          AMP binding
          kinase binding
          carbohydrate binding
          identical protein binding
          protein homodimerization activity
          metal ion binding
          fructose binding
          fructose-6-phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P251212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8398 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169199 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P251212 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P251212 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P251212 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P251212 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15527 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42261 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898976 - -
          Affymetrix Rat Gene 1.0 ST Array 10898980 - -
          Affymetrix Rat Gene 1.0 ST Array 10898972 - -
          Affymetrix Rat Gene 1.0 ST Array 10898977 - -
          Affymetrix Rat Gene 1.0 ST Array 10898982 - -
          Affymetrix Rat Gene 1.0 ST Array 10898986 - -
          Affymetrix Rat Gene 1.0 ST Array 10898973 - -
          Affymetrix Rat Gene 1.0 ST Array 10898987 - -
          Affymetrix Rat Gene 1.0 ST Array 10898975 - -
          Affymetrix Rat Gene 1.0 ST Array 10898979 - -
          Affymetrix Rat Gene 1.0 ST Array 10898984 - -
          Affymetrix Rat Gene 1.0 ST Array 10898985 - -
          Affymetrix Rat Gene 1.0 ST Array 10898989 - -
          Affymetrix Rat Gene 1.0 ST Array 10898981 - -
          Affymetrix Rat Gene 1.0 ST Array 10898988 - -
          Affymetrix Rat Gene 1.0 ST Array 10898970 - -
          Affymetrix Rat Gene 1.0 ST Array 10898983 - -
          Affymetrix Rat Gene 1.0 ST Array 10898971 - -
          Affymetrix Rat Gene 1.0 ST Array 10898974 - -
          Affymetrix Rat Gene 1.0 ST Array 10898990 - -
          Affymetrix Rat Gene 1.0 ST Array 10898969 - -
          Affymetrix Rat Gene 1.0 ST Array 10898978 - -
          Affymetrix Rat Gene 1.1 ST Array 10898968 - -
          Rat Genome 230 2.0 Array 1384632_PM_at - -
          Rat Genome 230 2.0 Array 1384632_at - -
          Rat Genome 230 2.0 Array 1367864_at - -
          Rat Genome 230 2.0 Array 1386961_PM_at - -
          Rat Genome 230 2.0 Array 1367864_PM_at - -
          Rat Genome 230 2.0 Array 1386961_at - -
          Affymetrix HT Rat Focus Array 1386961_PM_at - -
          Affymetrix HT Rat Focus Array 1386961_at - -
          Affymetrix HT Rat Focus Array 1384632_at - -
          Affymetrix HT Rat Focus Array 1384632_PM_at - -
          Affymetrix HT Rat Focus Array 1367864_at - -
          Affymetrix HT Rat Focus Array 1367864_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700013586.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013597.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013599.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013582.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013601.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013604.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013575.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013578.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013584.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013587.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013596.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013574.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013581.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013585.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013591.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013594.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013595.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013598.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013602.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013576.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013588.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013592.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013603.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013583.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013589.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013577.rn.1 TC0700001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013580.rn.1 TC0700001766.rn.1 RTA TC
          Rat Expression Set 230 1367864_PM_at - -
          Rat Expression Set 230 1386961_PM_at - -
          Rat Expression Set 230 1386961_at - -
          Rat Expression Set 230 1367864_at - -
          Rat Expression Set 230 1384632_PM_at - -
          Rat Expression Set 230 1384632_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898968 - -
          Rat Genome U34 Set D21869_s_at - -
          Rat Genome U34 Set U25651_at - -
          Rat Genome U34 Set rc_AI071717_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20713392 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364760 - -
          RatRef-12 Expression BeadChip ILMN_68863 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline