Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581864_m1
          See other CLNS1A GE Assays ›
          Gene Symbol:
          Clns1a
          Gene Name:
          chloride channel, nucleotide-sensitive, 1A
          Gene Aliases:
          Clci, Clcni, Icln, pICln
          Chromosome Location:
          Chr.1: 162676246 - 162696549 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581864_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clns1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031719.1 NP_113907.1 4 - 5 504 115
          XM_006229792.2 XP_006229854.1 4 - 5 567 115
          GenBank mRNA BC060555.1 - 4 - 5 548 6918578 115
          D13985.1 - 4 - 5 574 115
          D17698.1 - 4 - 5 487 115
          FQ212666.1 - 2 - 3 186 115
          FQ232880.1 - 2 - 3 186 115
          L26450.1 - 4 - 5 504 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002084.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clns1a

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal snRNP assembly
          chloride transport
          cell volume homeostasis
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P235815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21175 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87579 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P235815 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P235815 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P235815 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P235815 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17111 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41075 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10708825 - -
          Affymetrix Rat Gene 1.0 ST Array 10708831 - -
          Affymetrix Rat Gene 1.0 ST Array 10708826 - -
          Affymetrix Rat Gene 1.0 ST Array 10708828 - -
          Affymetrix Rat Gene 1.0 ST Array 10708827 - -
          Affymetrix Rat Gene 1.0 ST Array 10708829 - -
          Affymetrix Rat Gene 1.0 ST Array 10708830 - -
          Affymetrix Rat Gene 1.1 ST Array 10708824 - -
          Rat Genome 230 2.0 Array 1367772_at - -
          Rat Genome 230 2.0 Array 1367772_PM_at - -
          Affymetrix HT Rat Focus Array 1367772_at - -
          Affymetrix HT Rat Focus Array 1367772_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100016014.rn.1 TC0100002084.rn.1 RTA TC
          Rat Expression Set 230 1367772_PM_at - -
          Rat Expression Set 230 1367772_at - -
          Affymetrix Rat Gene 1.0 ST Array 10708824 - -
          Rat Genome U34 Set D13985_at - -
          Rat Genome U34 Set D13985_g_at - -
          Rat Genome U34 Set D13985_g_at - -
          Rat Genome U34 Set D13985_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22142431 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366686 - -
          RatRef-12 Expression BeadChip ILMN_60865 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline