Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581870_m1
          See other HTRA1 GE Assays ›
          Gene Symbol:
          Htra1
          Gene Name:
          HtrA serine peptidase 1
          Gene Aliases:
          Prss11
          Chromosome Location:
          Chr.1: 201499067 - 201548508 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581870_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htra1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031721.1 NP_113909.1 3 - 4 821 66
          GenBank mRNA AF179370.1 - 3 - 4 821 66
          BC081767.1 - 3 - 4 846 7121959 66
          FQ228942.1 - 2 - 3 299 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002705.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Htra1

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          proteolysis
          negative regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of BMP signaling pathway
          positive regulation of epithelial cell proliferation
          negative regulation of defense response to virus
          dentinogenesis
          serine-type endopeptidase activity
          insulin-like growth factor binding
          serine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P784614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152568 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171142 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115304 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P784614 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P784614 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P784614 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P784614 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700254054 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29954 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36594 - -
          Agilent Rat cDNA Microarray (G4105A) 700254054 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10711574 - -
          Affymetrix Rat Gene 1.0 ST Array 10711569 - -
          Affymetrix Rat Gene 1.0 ST Array 10711570 - -
          Affymetrix Rat Gene 1.0 ST Array 10711571 - -
          Affymetrix Rat Gene 1.0 ST Array 10711572 - -
          Affymetrix Rat Gene 1.0 ST Array 10711568 - -
          Affymetrix Rat Gene 1.0 ST Array 10711567 - -
          Affymetrix Rat Gene 1.0 ST Array 10711573 - -
          Affymetrix Rat Gene 1.1 ST Array 10711566 - -
          Rat Genome 230 2.0 Array 1386884_PM_at - -
          Rat Genome 230 2.0 Array 1386884_at - -
          Affymetrix HT Rat Focus Array 1386884_at - -
          Affymetrix HT Rat Focus Array 1386884_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100021281.rn.1 TC0100002705.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100021280.rn.1 TC0100002705.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100021273.rn.1 TC0100002705.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100021275.rn.1 TC0100002705.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100021279.rn.1 TC0100002705.rn.1 RTA TC
          Rat Expression Set 230 1386884_at - -
          Rat Expression Set 230 1386884_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10711566 - -
          Rat Genome U34 Set rc_AI013705_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21532821 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349449 - -
          RatRef-12 Expression BeadChip ILMN_64064 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline