Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581931_m1
          See other PPP5C GE Assays ›
          Gene Symbol:
          Ppp5c
          Gene Name:
          protein phosphatase 5, catalytic subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 78944054 - 78968516 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581931_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp5c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031729.1 NP_113917.1 2 - 3 372 77
          XM_006228465.1 XP_006228527.1 2 - 3 618 77
          GenBank mRNA BC078786.1 - 2 - 3 399 7126555 77
          U12203.1 - 2 - 3 300 77
          X77237.1 - 2 - 3 372 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004886.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp5c

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          protein dephosphorylation
          signal transduction
          response to lead ion
          dephosphorylation
          histone dephosphorylation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to morphine
          protein oligomerization
          protein heterooligomerization
          negative regulation of cell death
          cellular response to hydrogen peroxide
          cellular response to cadmium ion
          negative regulation of neuron death
          positive regulation of glucocorticoid receptor signaling pathway
          G-protein alpha-subunit binding
          RNA binding
          phosphoprotein phosphatase activity
          signal transducer activity
          microtubule binding
          heat shock protein binding
          identical protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59702 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52532 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69849 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409797 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409797 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P409797 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409797 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P168102 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700506927 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700783869 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28180 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54140 - -
          Agilent Rat cDNA Microarray (G4105A) 700506927 - -
          Agilent Rat cDNA Microarray (G4105A) 700783869 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719310 - -
          Affymetrix Rat Gene 1.0 ST Array 10719313 - -
          Affymetrix Rat Gene 1.0 ST Array 10719311 - -
          Affymetrix Rat Gene 1.0 ST Array 10719319 - -
          Affymetrix Rat Gene 1.0 ST Array 10719314 - -
          Affymetrix Rat Gene 1.0 ST Array 10719317 - -
          Affymetrix Rat Gene 1.0 ST Array 10719312 - -
          Affymetrix Rat Gene 1.0 ST Array 10719318 - -
          Affymetrix Rat Gene 1.0 ST Array 10719315 - -
          Affymetrix Rat Gene 1.0 ST Array 10719322 - -
          Affymetrix Rat Gene 1.0 ST Array 10719320 - -
          Affymetrix Rat Gene 1.0 ST Array 10719321 - -
          Affymetrix Rat Gene 1.0 ST Array 10719316 - -
          Affymetrix Rat Gene 1.1 ST Array 10719309 - -
          Rat Genome 230 2.0 Array 1397023_at - -
          Rat Genome 230 2.0 Array 1397023_PM_at - -
          Rat Genome 230 2.0 Array 1398242_at - -
          Rat Genome 230 2.0 Array 1398242_PM_at - -
          Affymetrix HT Rat Focus Array 1398242_PM_at - -
          Affymetrix HT Rat Focus Array 1398242_at - -
          Affymetrix Rat Clariom S Assay PSR0100038568.rn.1 TC0100004886.rn.1 RTA TC
          Rat Expression Set 230 1398242_at - -
          Rat Expression Set 230 1398242_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719309 - -
          Rat Genome U34 Set X77237_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21437327 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351857 - -
          RatRef-12 Expression BeadChip ILMN_55051 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline