Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581971_m1
          See other SLC27A2 GE Assays ›
          Gene Symbol:
          Slc27a2
          Gene Name:
          solute carrier family 27 (fatty acid transporter), member 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 119014620 - 119052531 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581971_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc27a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031736.1 NP_113924.1 8 - 9 2351 84
          GenBank mRNA BC081766.1 - 8 - 9 1688 7106977 84
          D85100.1 - 8 - 9 2351 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001642.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc27a2

          Panther Classification:

          Molecular Function -

          transporter secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          very long-chain fatty acid metabolic process
          fatty acid alpha-oxidation
          long-chain fatty acid metabolic process
          fatty acid beta-oxidation
          bile acid biosynthetic process
          fatty acid transport
          very long-chain fatty acid catabolic process
          long-chain fatty acid import
          methyl-branched fatty acid metabolic process
          long-chain fatty acid-CoA ligase activity
          receptor binding
          protein binding
          ATP binding
          fatty acid transporter activity
          enzyme binding
          very long-chain fatty acid-CoA ligase activity
          phytanate-CoA ligase activity
          pristanate-CoA ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P103632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77341 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124321 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96693 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12655 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12655 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12655 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12655 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P997755 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700625164 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73283 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59245 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38130 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44340 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88959 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12197 - -
          Agilent Rat cDNA Microarray (G4105A) 700458952 - -
          Agilent Rat cDNA Microarray (G4105A) 700625164 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839444 - -
          Affymetrix Rat Gene 1.0 ST Array 10839448 - -
          Affymetrix Rat Gene 1.0 ST Array 10839447 - -
          Affymetrix Rat Gene 1.0 ST Array 10839450 - -
          Affymetrix Rat Gene 1.0 ST Array 10839446 - -
          Affymetrix Rat Gene 1.0 ST Array 10839449 - -
          Affymetrix Rat Gene 1.0 ST Array 10839453 - -
          Affymetrix Rat Gene 1.0 ST Array 10839454 - -
          Affymetrix Rat Gene 1.0 ST Array 10839445 - -
          Affymetrix Rat Gene 1.0 ST Array 10839451 - -
          Affymetrix Rat Gene 1.0 ST Array 10839452 - -
          Affymetrix Rat Gene 1.1 ST Array 10839443 - -
          Rat Genome 230 2.0 Array 1368150_PM_at - -
          Rat Genome 230 2.0 Array 1368150_at - -
          Affymetrix HT Rat Focus Array 1368150_PM_at - -
          Affymetrix HT Rat Focus Array 1368150_at - -
          Affymetrix Rat Clariom S Assay PSR0300011556.rn.1 TC0300001642.rn.1 RTA TC
          Rat Expression Set 230 1368150_at - -
          Rat Expression Set 230 1368150_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839443 - -
          Rat Genome U34 Set D85100_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22323350 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline