Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582000_m1
          See other SLC2A5 GE Assays ›
          Gene Symbol:
          Slc2a5
          Gene Name:
          solute carrier family 2 (facilitated glucose/fructose transporter), member 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 167142182 - 167174203 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582000_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc2a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031741.1 NP_113929.1 5 - 6 607 57
          GenBank mRNA BC076378.1 - 5 - 6 592 57
          BC161843.1 - 5 - 6 594 7127817 57
          D13871.1 - 5 - 6 607 57
          D28562.1 - 5 - 6 566 57
          L05195.1 - 5 - 6 579 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001955.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc2a5

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose transport
          glucose transport
          transmembrane transport
          cellular response to fructose stimulus
          fructose transmembrane transporter activity
          glucose transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44804 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55363 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022155 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022155 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022155 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P520497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52332 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1605 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38166 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10874136 - -
          Affymetrix Rat Gene 1.0 ST Array 10874133 - -
          Affymetrix Rat Gene 1.0 ST Array 10874132 - -
          Affymetrix Rat Gene 1.0 ST Array 10874131 - -
          Affymetrix Rat Gene 1.0 ST Array 10874128 - -
          Affymetrix Rat Gene 1.0 ST Array 10874138 - -
          Affymetrix Rat Gene 1.0 ST Array 10874137 - -
          Affymetrix Rat Gene 1.0 ST Array 10874135 - -
          Affymetrix Rat Gene 1.0 ST Array 10874134 - -
          Affymetrix Rat Gene 1.0 ST Array 10874139 - -
          Affymetrix Rat Gene 1.0 ST Array 10874140 - -
          Affymetrix Rat Gene 1.0 ST Array 10874129 - -
          Affymetrix Rat Gene 1.0 ST Array 10874127 - -
          Affymetrix Rat Gene 1.0 ST Array 10874130 - -
          Affymetrix Rat Gene 1.1 ST Array 10874126 - -
          Rat Genome 230 2.0 Array 1368460_PM_at - -
          Rat Genome 230 2.0 Array 1368460_at - -
          Affymetrix HT Rat Focus Array 1368460_PM_at - -
          Affymetrix HT Rat Focus Array 1368460_at - -
          Affymetrix Rat Clariom S Assay PSR0500014655.rn.1 TC0500001955.rn.1 RTA TC
          Rat Expression Set 230 1368460_PM_at - -
          Rat Expression Set 230 1368460_at - -
          Affymetrix Rat Gene 1.0 ST Array 10874126 - -
          Rat Genome U34 Set D28562_s_at - -
          Rat Genome U34 Set D13871_s_at - -
          Rat Genome U34 Set D13871_s_at - -
          Rat Genome U34 Set D28562_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20882596 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374014 - -
          RatRef-12 Expression BeadChip ILMN_50042 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0