Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582330_s1
          See other KCNS3 GE Assays ›
          Gene Symbol:
          Kcns3
          Gene Name:
          potassium voltage-gated channel, modifier subfamily S, member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 36764040 - 36819810 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582330_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcns3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031778.2 NP_113966.2 3 754 67
          GenBank mRNA AF029056.1 - 1 452 67
          Y17607.1 - 3 754 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcns3

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ion transmembrane transport
          protein homooligomerization
          potassium ion transmembrane transport
          delayed rectifier potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P416938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2273 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143804 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40826 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P416938 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P416938 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P416938 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P416938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78916 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102658 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80482 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20213 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10889191 - -
          Affymetrix Rat Gene 1.0 ST Array 10889193 - -
          Affymetrix Rat Gene 1.0 ST Array 10889192 - -
          Affymetrix Rat Gene 1.1 ST Array 10889190 - -
          Rat Genome 230 2.0 Array 1368751_PM_at - -
          Rat Genome 230 2.0 Array 1368751_at - -
          Affymetrix HT Rat Focus Array 1368751_at - -
          Affymetrix HT Rat Focus Array 1368751_PM_at - -
          Rat Expression Set 230 1368751_PM_at - -
          Rat Expression Set 230 1368751_at - -
          Affymetrix Rat Gene 1.0 ST Array 10889190 - -
          Rat Genome U34 Set Y17607_s_at - -
          Rat Genome U34 Set Y17607_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21453691 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354516 - -
          RatRef-12 Expression BeadChip ILMN_60844 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline