Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582409_m1
          See other HIPK3 GE Assays ›
          Gene Symbol:
          Hipk3
          Gene Name:
          homeodomain interacting protein kinase 3
          Gene Aliases:
          ANPK
          Chromosome Location:
          Chr.3: 94349778 - 94419191 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582409_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hipk3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031787.1 NP_113975.1 6 - 7 2084 107
          XM_006234658.2 XP_006234720.1 6 - 7 2225 107
          XM_008762072.1 XP_008760294.1 6 - 7 2227 107
          GenBank mRNA AF036959.1 - 6 - 7 2084 107
          BC100068.1 - 6 - 7 1734 7111326 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003783.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hipk3

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          mRNA transcription
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          negative regulation of apoptotic process
          negative regulation of JUN kinase activity
          DNA binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P800949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3827 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127012 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45785 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52224 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P800949 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P800949 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P800949 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P800949 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701917014 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21694 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37099 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24671 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36601 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88393 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3218 - -
          Agilent Rat cDNA Microarray (G4105A) 701917014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10847911 - -
          Affymetrix Rat Gene 1.0 ST Array 10847916 - -
          Affymetrix Rat Gene 1.0 ST Array 10847920 - -
          Affymetrix Rat Gene 1.0 ST Array 10847917 - -
          Affymetrix Rat Gene 1.0 ST Array 10847918 - -
          Affymetrix Rat Gene 1.0 ST Array 10847908 - -
          Affymetrix Rat Gene 1.0 ST Array 10847910 - -
          Affymetrix Rat Gene 1.0 ST Array 10847904 - -
          Affymetrix Rat Gene 1.0 ST Array 10847913 - -
          Affymetrix Rat Gene 1.0 ST Array 10847914 - -
          Affymetrix Rat Gene 1.0 ST Array 10847919 - -
          Affymetrix Rat Gene 1.0 ST Array 10847905 - -
          Affymetrix Rat Gene 1.0 ST Array 10847906 - -
          Affymetrix Rat Gene 1.0 ST Array 10847915 - -
          Affymetrix Rat Gene 1.0 ST Array 10847907 - -
          Affymetrix Rat Gene 1.0 ST Array 10847921 - -
          Affymetrix Rat Gene 1.0 ST Array 10847912 - -
          Affymetrix Rat Gene 1.1 ST Array 10847903 - -
          Rat Genome 230 2.0 Array 1369352_at - -
          Rat Genome 230 2.0 Array 1375274_PM_at - -
          Rat Genome 230 2.0 Array 1369352_PM_at - -
          Rat Genome 230 2.0 Array 1377127_at - -
          Rat Genome 230 2.0 Array 1377127_PM_at - -
          Rat Genome 230 2.0 Array 1375274_at - -
          Affymetrix HT Rat Focus Array 1375274_PM_at - -
          Affymetrix HT Rat Focus Array 1369352_PM_at - -
          Affymetrix HT Rat Focus Array 1375274_at - -
          Affymetrix HT Rat Focus Array 1369352_at - -
          Affymetrix Rat Clariom S Assay PSR0300028867.rn.1 TC0300003783.rn.1 RTA TC
          Rat Expression Set 230 1377127_PM_at - -
          Rat Expression Set 230 1375274_PM_at - -
          Rat Expression Set 230 1377127_at - -
          Rat Expression Set 230 1369352_at - -
          Rat Expression Set 230 1369352_PM_at - -
          Rat Expression Set 230 1375274_at - -
          Affymetrix Rat Gene 1.0 ST Array 10847903 - -
          Rat Genome U34 Set rc_AA866455_at - -
          Rat Genome U34 Set rc_AA866455_g_at - -
          Rat Genome U34 Set AF036959_at - -
          Rat Genome U34 Set rc_AA925912_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22114785 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369988 - -
          RatRef-12 Expression BeadChip ILMN_58063 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline