Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582449_m1
          See other CIT GE Assays ›
          Gene Symbol:
          Cit
          Gene Name:
          citron rho-interacting serine/threonine kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 46334669 - 46494155 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582449_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cit
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029911.1 NP_001025082.1 17 - 18 2316 64
          XM_006249469.2 XP_006249531.1 18 - 19 2294 64
          XM_006249470.2 XP_006249532.1 18 - 19 2294 64
          XM_006249471.2 XP_006249533.1 18 - 19 2294 64
          XM_006249472.2 XP_006249534.1 18 - 19 2294 64
          XM_006249477.2 XP_006249539.1 8 - 9 1482 64
          XM_008769269.1 XP_008767491.1 18 - 19 2294 64
          XM_008769270.1 XP_008767492.1 18 - 19 2250 64
          XM_008769271.1 XP_008767493.1 17 - 18 2320 64
          GenBank mRNA AF039218.1 - 8 - 9 1420 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001731.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cit

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          G2/M transition of mitotic cell cycle
          protein phosphorylation
          Golgi organization
          mitotic nuclear division
          metaphase/anaphase transition of mitotic cell cycle
          spermatogenesis
          regulation of actin polymerization or depolymerization
          phosphorylation
          dendrite development
          positive regulation of cytokinesis
          intracellular signal transduction
          generation of neurons
          negative regulation of dendrite morphogenesis
          regulation of catalytic activity
          regulation of cell cycle
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          Rho GTPase binding
          PDZ domain binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890230 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119161 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16726 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173222 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129498 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129498 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15913 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P477879 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P990522 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701905953 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39616 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79944 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35484 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68359 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90580 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40069 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60579 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103833 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3930 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58458 - -
          Agilent Rat cDNA Microarray (G4105A) 701905953 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762545 - -
          Affymetrix Rat Gene 1.0 ST Array 10762558 - -
          Affymetrix Rat Gene 1.0 ST Array 10762542 - -
          Affymetrix Rat Gene 1.0 ST Array 10762582 - -
          Affymetrix Rat Gene 1.0 ST Array 10762540 - -
          Affymetrix Rat Gene 1.0 ST Array 10762541 - -
          Affymetrix Rat Gene 1.0 ST Array 10762572 - -
          Affymetrix Rat Gene 1.0 ST Array 10762543 - -
          Affymetrix Rat Gene 1.0 ST Array 10762568 - -
          Affymetrix Rat Gene 1.0 ST Array 10762556 - -
          Affymetrix Rat Gene 1.0 ST Array 10762580 - -
          Affymetrix Rat Gene 1.0 ST Array 10762553 - -
          Affymetrix Rat Gene 1.0 ST Array 10762539 - -
          Affymetrix Rat Gene 1.0 ST Array 10762557 - -
          Affymetrix Rat Gene 1.0 ST Array 10762562 - -
          Affymetrix Rat Gene 1.0 ST Array 10762587 - -
          Affymetrix Rat Gene 1.0 ST Array 10762551 - -
          Affymetrix Rat Gene 1.0 ST Array 10762570 - -
          Affymetrix Rat Gene 1.0 ST Array 10762569 - -
          Affymetrix Rat Gene 1.0 ST Array 10762555 - -
          Affymetrix Rat Gene 1.0 ST Array 10762563 - -
          Affymetrix Rat Gene 1.0 ST Array 10762571 - -
          Affymetrix Rat Gene 1.0 ST Array 10762576 - -
          Affymetrix Rat Gene 1.0 ST Array 10762573 - -
          Affymetrix Rat Gene 1.0 ST Array 10762586 - -
          Affymetrix Rat Gene 1.0 ST Array 10762544 - -
          Affymetrix Rat Gene 1.0 ST Array 10762550 - -
          Affymetrix Rat Gene 1.0 ST Array 10762547 - -
          Affymetrix Rat Gene 1.0 ST Array 10762577 - -
          Affymetrix Rat Gene 1.0 ST Array 10762566 - -
          Affymetrix Rat Gene 1.0 ST Array 10762559 - -
          Affymetrix Rat Gene 1.0 ST Array 10762552 - -
          Affymetrix Rat Gene 1.0 ST Array 10762579 - -
          Affymetrix Rat Gene 1.0 ST Array 10762583 - -
          Affymetrix Rat Gene 1.0 ST Array 10762578 - -
          Affymetrix Rat Gene 1.0 ST Array 10762565 - -
          Affymetrix Rat Gene 1.0 ST Array 10762585 - -
          Affymetrix Rat Gene 1.0 ST Array 10762554 - -
          Affymetrix Rat Gene 1.0 ST Array 10762560 - -
          Affymetrix Rat Gene 1.0 ST Array 10762538 - -
          Affymetrix Rat Gene 1.0 ST Array 10762548 - -
          Affymetrix Rat Gene 1.0 ST Array 10762584 - -
          Affymetrix Rat Gene 1.0 ST Array 10762546 - -
          Affymetrix Rat Gene 1.0 ST Array 10762588 - -
          Affymetrix Rat Gene 1.0 ST Array 10762564 - -
          Affymetrix Rat Gene 1.0 ST Array 10762575 - -
          Affymetrix Rat Gene 1.0 ST Array 10762574 - -
          Affymetrix Rat Gene 1.0 ST Array 10762581 - -
          Affymetrix Rat Gene 1.0 ST Array 10762567 - -
          Affymetrix Rat Gene 1.0 ST Array 10762549 - -
          Affymetrix Rat Gene 1.0 ST Array 10762561 - -
          Affymetrix Rat Gene 1.1 ST Array 10762537 - -
          Rat Genome 230 2.0 Array 1389137_at - -
          Rat Genome 230 2.0 Array 1389137_PM_at - -
          Rat Genome 230 2.0 Array 1387381_at - -
          Rat Genome 230 2.0 Array 1387381_PM_at - -
          Affymetrix HT Rat Focus Array 1389137_PM_at - -
          Affymetrix HT Rat Focus Array 1389137_at - -
          Affymetrix HT Rat Focus Array 1387381_PM_at - -
          Affymetrix HT Rat Focus Array 1387381_at - -
          Affymetrix Rat Clariom S Assay PSR1200014706.rn.1 TC1200001731.rn.1 RTA TC
          Rat Expression Set 230 1389137_PM_at - -
          Rat Expression Set 230 1387381_PM_at - -
          Rat Expression Set 230 1389137_at - -
          Rat Expression Set 230 1387381_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762537 - -
          Rat Genome U34 Set AF039218_at - -
          Rat Genome U34 Set rc_AI059345_at - -
          Rat Genome U34 Set rc_AA955912_at - -
          Rat Genome U34 Set rc_AI059948_s_at - -
          Rat Genome U34 Set rc_AI059948_s_at - -
          Rat Genome U34 Set AF039218_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21443557 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364883 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline