Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582574_m1
          See other CAPN6 GE Assays ›
          Gene Symbol:
          Capn6
          Gene Name:
          calpain 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 115049113 - 115073824 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582574_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Capn6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031808.1 NP_113996.1 2 - 3 330 70
          GenBank mRNA AF067793.1 - 2 - 3 330 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002283.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Capn6

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule bundle formation
          proteolysis
          regulation of cytoskeleton organization
          calcium-dependent cysteine-type endopeptidase activity
          microtubule binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P121643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156828 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95218 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121643 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121643 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P121643 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P279479 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38910 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10932761 - -
          Affymetrix Rat Gene 1.0 ST Array 10932760 - -
          Affymetrix Rat Gene 1.0 ST Array 10932767 - -
          Affymetrix Rat Gene 1.0 ST Array 10932769 - -
          Affymetrix Rat Gene 1.0 ST Array 10932763 - -
          Affymetrix Rat Gene 1.0 ST Array 10932770 - -
          Affymetrix Rat Gene 1.0 ST Array 10932771 - -
          Affymetrix Rat Gene 1.0 ST Array 10932768 - -
          Affymetrix Rat Gene 1.0 ST Array 10932772 - -
          Affymetrix Rat Gene 1.0 ST Array 10932764 - -
          Affymetrix Rat Gene 1.0 ST Array 10932766 - -
          Affymetrix Rat Gene 1.0 ST Array 10932765 - -
          Affymetrix Rat Gene 1.0 ST Array 10932762 - -
          Affymetrix Rat Gene 1.1 ST Array 10932759 - -
          Rat Genome 230 2.0 Array 1368131_PM_at - -
          Rat Genome 230 2.0 Array 1368131_at - -
          Affymetrix HT Rat Focus Array 1368131_PM_at - -
          Affymetrix HT Rat Focus Array 1368131_at - -
          Affymetrix Rat Clariom S Assay PSR0X00014331.rn.1 TC0X00002283.rn.1 RTA TC
          Rat Expression Set 230 1368131_at - -
          Rat Expression Set 230 1368131_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10932759 - -
          Rat Genome U34 Set rc_AI171796_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21610996 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376248 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline