Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582649_m1
          See other GIT1 GE Assays ›
          Gene Symbol:
          Git1
          Gene Name:
          G protein-coupled receptor kinase interacting ArfGAP 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 62656000 - 62664467 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582649_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Git1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031814.1 NP_114002.1 6 - 7 820 95
          GenBank mRNA AF085693.1 - 6 - 7 820 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003257.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Git1

          Gene Ontology Categories:

          Function(s) Process(es)

          immunological synapse formation
          regulation of G-protein coupled receptor protein signaling pathway
          regulation of cytokinesis
          positive regulation of GTPase activity
          ephrin receptor signaling pathway
          dendritic spine development
          GTPase activator activity
          protein binding
          zinc ion binding
          small GTPase binding
          protein complex binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P081884 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119266 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12692 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081884 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12692 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081884 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12692 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P081884 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12692 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700485536 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101910 - -
          Agilent Rat cDNA Microarray (G4105A) 700485536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10745399 - -
          Affymetrix Rat Gene 1.0 ST Array 10745394 - -
          Affymetrix Rat Gene 1.0 ST Array 10745389 - -
          Affymetrix Rat Gene 1.0 ST Array 10745398 - -
          Affymetrix Rat Gene 1.0 ST Array 10745392 - -
          Affymetrix Rat Gene 1.0 ST Array 10745388 - -
          Affymetrix Rat Gene 1.0 ST Array 10745396 - -
          Affymetrix Rat Gene 1.0 ST Array 10745403 - -
          Affymetrix Rat Gene 1.0 ST Array 10745404 - -
          Affymetrix Rat Gene 1.0 ST Array 10745397 - -
          Affymetrix Rat Gene 1.0 ST Array 10745391 - -
          Affymetrix Rat Gene 1.0 ST Array 10745395 - -
          Affymetrix Rat Gene 1.0 ST Array 10745405 - -
          Affymetrix Rat Gene 1.0 ST Array 10745400 - -
          Affymetrix Rat Gene 1.0 ST Array 10745407 - -
          Affymetrix Rat Gene 1.0 ST Array 10745401 - -
          Affymetrix Rat Gene 1.0 ST Array 10745390 - -
          Affymetrix Rat Gene 1.0 ST Array 10745406 - -
          Affymetrix Rat Gene 1.0 ST Array 10745393 - -
          Affymetrix Rat Gene 1.1 ST Array 10745387 - -
          Rat Genome 230 2.0 Array 1370015_PM_at - -
          Rat Genome 230 2.0 Array 1370015_at - -
          Affymetrix HT Rat Focus Array 1370015_PM_at - -
          Affymetrix HT Rat Focus Array 1370015_at - -
          Affymetrix Rat Clariom S Assay PSR1000029190.rn.1 TC1000003257.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029197.rn.1 TC1000003257.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029199.rn.1 TC1000003257.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029192.rn.1 TC1000003257.rn.1 RTA TC
          Rat Expression Set 230 1370015_PM_at - -
          Rat Expression Set 230 1370015_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745387 - -
          Rat Genome U34 Set AF085693_at - -
          Rat Genome U34 Set AF085693_g_at - -
          Rat Genome U34 Set rc_AI009086_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20945417 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362046 - -
          RatRef-12 Expression BeadChip ILMN_63588 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline