Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582667_m1
          See other CLIC4 GE Assays ›
          Gene Symbol:
          Clic4
          Gene Name:
          chloride intracellular channel 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 153568937 - 153625669 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582667_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clic4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031818.1 NP_114006.1 4 - 5 412 76
          GenBank mRNA AF104119.1 - 4 - 5 412 76
          FQ222737.1 - 522 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003831.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clic4

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          endothelial cell morphogenesis
          chloride transport
          apoptotic process
          vacuolar acidification
          fertilization
          keratinocyte differentiation
          negative regulation of cell migration
          regulation of ion transmembrane transport
          multicellular organism growth
          branching morphogenesis of an epithelial tube
          retina vasculature morphogenesis in camera-type eye
          cellular response to calcium ion
          chloride transmembrane transport
          voltage-gated ion channel activity
          chloride channel activity
          protein binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104429 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54296 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118560 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88537 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119853 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054538 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054538 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054538 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P105831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94735 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37361 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60209 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63613 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880585 - -
          Affymetrix Rat Gene 1.0 ST Array 10880587 - -
          Affymetrix Rat Gene 1.0 ST Array 10880586 - -
          Affymetrix Rat Gene 1.0 ST Array 10880584 - -
          Affymetrix Rat Gene 1.0 ST Array 10880589 - -
          Affymetrix Rat Gene 1.0 ST Array 10880588 - -
          Affymetrix Rat Gene 1.1 ST Array 10880583 - -
          Rat Genome 230 2.0 Array 1377765_at - -
          Rat Genome 230 2.0 Array 1377765_PM_at - -
          Rat Genome 230 2.0 Array 1387420_at - -
          Rat Genome 230 2.0 Array 1387420_PM_at - -
          Affymetrix HT Rat Focus Array 1387420_at - -
          Affymetrix HT Rat Focus Array 1377765_at - -
          Affymetrix HT Rat Focus Array 1387420_PM_at - -
          Affymetrix HT Rat Focus Array 1377765_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500028770.rn.1 TC0500003831.rn.1 RTA TC
          Rat Expression Set 230 1387420_at - -
          Rat Expression Set 230 1387420_PM_at - -
          Rat Expression Set 230 1377765_at - -
          Rat Expression Set 230 1377765_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880583 - -
          Rat Genome U34 Set rc_AI030141_at - -
          Rat Genome U34 Set rc_AA819415_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21711111 - -
          ABI Rat Genome Survey Microarray 22333533 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363135 - -
          RatRef-12 Expression BeadChip ILMN_69999 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline